Skip to Content
Merck
All Photos(2)

Key Documents

45696

Supelco

Triazophos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N3O3PS
CAS Number:
Molecular Weight:
313.31
Beilstein:
682554
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1ncn(n1)-c2ccccc2

InChI

1S/C12H16N3O3PS/c1-3-16-19(20,17-4-2)18-12-13-10-15(14-12)11-8-6-5-7-9-11/h5-10H,3-4H2,1-2H3

InChI key

AMFGTOFWMRQMEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mubeena Akhtar et al.
Colloids and surfaces. B, Biointerfaces, 69(1), 63-70 (2008-12-19)
The sorption efficiency of chickpea husk of black gram variety (BGH), for the removal of organophosphorous pesticides (OPPs), i.e. triazophos (TAP) and methyl parathion (MP) from aqueous media has been investigated. Optimization of operating sorption parameters, i.e. particle size, sorbent
Yan-Yuan Bao et al.
Journal of insect physiology, 56(9), 1087-1094 (2010-03-13)
The widespread use of insecticides has caused the resurgence of the brown planthopper, Nilaparvata lugens, in Asia. In this study, we investigated an organo-phosphorous insecticide, triazophos, and its ability to induce gene expression variation in female N. lugens nymphs just
Lin-Quan Ge et al.
Journal of proteome research, 10(10), 4597-4612 (2011-08-02)
The brown planthopper, Nilaparvata lugens (Stål) (Hemiptera: Delphacidae), is a classic example of a resurgent pest induced by insecticides. It has been demonstrated that triazophos treatment causes an increase in the content of male accessory gland proteins (Acps) that can
Shaonan Li et al.
Journal of environmental sciences (China), 23(5), 852-859 (2011-07-28)
In vivo activity of cholinesterase (ChE) in Daphnia magna was measured at different time points during 21-day exposure to triazophos and chlorpyrifos ranging from 0.05 to 2.50 microg/L and 0.01 to 2.00 microg/L, respectively. For exposure to triazophos, ChE was
Hongcui Liu et al.
Ecotoxicology and environmental safety, 80, 118-125 (2012-03-23)
For investigating relationship between activity of cholinesterase (ChE) and ambient concentration of anticholinesterases, Daphnia magna had been exposed for 21 day to sub-lethal concentrations, i.e. 1/6 EC(50), 1/36 EC(50), and 1/216 EC(50), of either triazophos or chlorpyrifos. Samples were taken

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service