Skip to Content
Merck
All Photos(1)

Key Documents

45643

Supelco

Propineb

PESTANAL®, analytical standard

Synonym(s):

Zinc propylenebis(dithiocarbamate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C5H8N2S4Zn]n
CAS Number:
Beilstein:
2360676
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

technical mixture

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

mol wt

Mw 289.78 g/mol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1CNC(=S)S[Zn]SC(=S)N1

InChI

1S/C5H10N2S4.Zn/c1-3(7-5(10)11)2-6-4(8)9;/h3H,2H2,1H3,(H2,6,8,9)(H2,7,10,11);/q;+2/p-2

InChI key

KKMLIVYBGSAJPM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Propineb belongs to the dithiocarbamates (DTCs) class of fungicides, widely used in agriculture and horticulture. It has an additional advantage of low production cost, broad-spectrum of chemical and biological reactivity.

Application

Propineb may be used as an analytical reference standard for the quantification of the analyte in environmental samples and vegetable samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kelly Flampouri et al.
Talanta, 80(5), 1799-1804 (2010-02-16)
Two of the most important categories of pesticides used in agricultural practice are organophosphates and dithiocarbamates. Their extensive and inappropriate use has rendered their reliable monitoring at trace levels more and more necessary. This study presents the construction of a
M Yaldiz et al.
Clinical and experimental obstetrics & gynecology, 27(3-4), 235-237 (2001-02-24)
The dithiocarbamates are known to cause dermatitis, conjunctivitis, rhinitis, pharyngitis and bronchitis in humans. The experimental group received Propineb (obtained from Bayer) concentrations of 400 ppm in distilled water five days a week (treatment time three weeks) administered orally by
Marina Marinovich et al.
Chemical research in toxicology, 15(1), 26-32 (2002-01-22)
Dithiocarbamates (DTCs) are used mainly in agriculture as pesticides and as alcohol deterrent drugs. Neurological complications as well as movement disorders characterized by plastic rigidity, muscle twitch and paralysis are the prevailing symptoms in chronically exposed animals and humans. We
A T Wojdyła
Communications in agricultural and applied biological sciences, 69(4), 705-715 (2005-03-11)
Chitosan, (Biochikol 020 PC) a potential elicitor of plant defence and also an active inhibitor of fungal growth was used in experiments. The compound was used at concentrations 0.01 to 0.2% as a plant spray for rose protection against Sphaerotheca
Barbara Viviani et al.
Toxicology letters, 182(1-3), 63-68 (2008-09-30)
Neurological complications as well as movement disorders are relevant symptoms in animals and humans chronically exposed to dithiocarbamates. Using rat pheochromocytoma cells differentiated by NGF (PC12), we investigated whether propineb affects acetylcholine (Ach) release and the molecular mechanisms involved. Propineb

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service