Skip to Content
Merck
All Photos(1)

Key Documents

45506

Supelco

Fluorodifen

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H7F3N2O5
CAS Number:
Molecular Weight:
328.20
Beilstein:
2018474
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[O-][N+](=O)c1ccc(Oc2ccc(cc2[N+]([O-])=O)C(F)(F)F)cc1

InChI

1S/C13H7F3N2O5/c14-13(15,16)8-1-6-12(11(7-8)18(21)22)23-10-4-2-9(3-5-10)17(19)20/h1-7H

InChI key

HHMCAJWVGYGUEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniele Del Buono et al.
Journal of agricultural and food chemistry, 59(22), 12109-12115 (2011-10-18)
A study was carried out to compare the effects of treating wheat (Triticum aestivum) and Italian ryegrass (Lolium multiflorum) with atrazine and fluorodifen. The herbicides interfered with photosynthesis and dark respiration, depending on the species. Atrazine decreased photosynthesis in both
Daniele Del Buono et al.
Journal of agricultural and food chemistry, 57(17), 7924-7930 (2009-08-08)
The effect of treatments with four herbicides and a safener on the activity of triosephosphate isomerase (TPI) extracted from shoots of Italian ryegrass was investigated. It was found that atrazine and fluorodifen, herbicides which interfere with photosynthesis, caused a decrease
S Pflugmacher et al.
Phytochemistry, 54(3), 267-273 (2000-06-28)
Soluble and microsomal glutathione S-transferase activities for five model xenobiotics (nitrobenzene derivatives), two pesticidal xenobiotics (atrazine and fluorodifen), and a natural substrate (cinnamic acid), were determined in 59 different plant species and four plant cell suspension cultures. These enzyme activities
Daniele Del Buono et al.
Journal of agricultural and food chemistry, 59(4), 1324-1329 (2011-02-04)
Many varieties of Italian ryegrass (Lolium multiflorum) show resistance to herbicides; while this ability was frequently attributed to alterations in the target sites of the herbicide's action of the plant or to an efficient oxidative metabolism, little attention has been
Irene Axarli et al.
Journal of molecular biology, 385(3), 984-1002 (2008-11-19)
Glutathione transferases (GSTs) from the tau class (GSTU) are unique to plants and have important roles in stress tolerance and the detoxification of herbicides in crops and weeds. A fluorodifen-induced GST isoezyme (GmGSTU4-4) belonging to the tau class was purified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service