Skip to Content
Merck
All Photos(1)

Key Documents

31713

Supelco

Fenhexamid

PESTANAL®, analytical standard

Synonym(s):

N-(2,3-Dichloro-4-hydroxyphenyl)-1-methylcyclohexanecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17Cl2NO2
CAS Number:
Molecular Weight:
302.20
Beilstein:
8996658
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(CCCCC1)C(=O)Nc2ccc(O)c(Cl)c2Cl

InChI

1S/C14H17Cl2NO2/c1-14(7-3-2-4-8-14)13(19)17-9-5-6-10(18)12(16)11(9)15/h5-6,18H,2-4,7-8H2,1H3,(H,17,19)

InChI key

VDLGAVXLJYLFDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenhexamid is an anti-botrytis fungicide widely used against a broad spectrum of fungi in agricultural crops. Its mode of action involves the inhibition of sterol biosynthesis.

Application

Fenhexamid may be used as a standard in the determination of fenhexamid in:
  • Fruits using enzyme-linked immunosorbent assay (ELISA).
  • Tomatoes, grape and wine samples using high-performance liquid chromatography coupled with ultraviolet detector and gas chromatography (GC) coupled with detectors such as nitrogen–phosphorus detector (NPD), electron capture detector (ECD) and ion trap mass spectrometry (ITMS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Kretschmer et al.
PLoS pathogens, 5(12), e1000696-e1000696 (2009-12-19)
The grey mould fungus Botrytis cinerea causes losses of commercially important fruits, vegetables and ornamentals worldwide. Fungicide treatments are effective for disease control, but bear the risk of resistance development. The major resistance mechanism in fungi is target protein modification
Kim Christopher Cohrs et al.
Fungal genetics and biology : FG & B, 100, 42-51 (2017-02-12)
The gray mold fungus Botrytis cinerea features a wide host range and causes severe economic losses, making it an important object for molecular research. Thus far, genetic modification of the fungus mainly is relied on two selection systems (nourseothricin and
Determination of fenhexamid residues in grape must, kiwifruit, and strawberry samples by enzyme-linked immunosorbent assay
Esteve-Turrillas.AF, et al.
Food Chemistry, 124(4), 1727-1733 (2011)
The hydroxyanilide fenhexamid, a new sterol biosynthesis inhibitor fungicide efficient against the plant pathogenic fungus Botryotinia fuckeliana (Botrytis cinerea)
Debieu D, et al.
Pest Management Science, 57(11), 1060-1067 (2001)
Facundo Rivera-Becerril et al.
The Science of the total environment, 577, 84-93 (2016-11-08)
Pesticide contamination of the environment can result from agricultural practices. Persistence of pesticide residues is a threat to the soil biota including plant roots and beneficial microorganisms, which support an important number of soil ecosystem services. Arbuscular mycorrhizal fungi (AMF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service