Skip to Content
Merck
All Photos(1)

Key Documents

45403

Supelco

Coumaphos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16ClO5PS
CAS Number:
Molecular Weight:
362.77
Beilstein:
327083
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OCC)Oc1ccc2C(C)=C(Cl)C(=O)Oc2c1

InChI

1S/C14H16ClO5PS/c1-4-17-21(22,18-5-2)20-10-6-7-11-9(3)13(15)14(16)19-12(11)8-10/h6-8H,4-5H2,1-3H3

InChI key

BXNANOICGRISHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharath V Sagar et al.
Tropical animal health and production, 52(2), 611-618 (2019-08-30)
The isolates of Rhipicephalus microplus collected from Madhya Pradesh (MP), Punjab (PJB) and Uttar Pradesh (UP) states of India were characterized using laboratory standardized adult immersion test (AIT) against macrocyclic lactone (ivermectin), synthetic pyrethroids (cypermethrin and deltamethrin), organophosphates (coumaphos and
Carmen Sara Hernández-Rodríguez et al.
Insects, 12(1) (2021-01-08)
Varroa destructor is an ectoparasitic mite causing devastating damages to honey bee colonies around the world. Its impact is considered a major factor contributing to the significant seasonal losses of colonies recorded every year. Beekeepers usually rely on a reduced
Emmanuel Pacia Hernandez et al.
Parasites & vectors, 11(1), 460-460 (2018-08-11)
Haemaphysalis longicornis is a tick of importance to health, as it serves as a vector of several pathogens, including Theileria orientalis, Babesia ovata, Rickettsia japonica and the severe fever with thrombocytopenia syndrome virus (SFTSV). Presently, the major method of control
Wenfu Mao et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(31), 12657-12662 (2011-07-22)
Although Apis mellifera, the western honey bee, has long encountered pesticides when foraging in agricultural fields, for two decades it has encountered pesticides in-hive in the form of acaricides to control Varroa destructor, a devastating parasitic mite. The pyrethroid tau-fluvalinate
Katja Schweikert et al.
Aquatic toxicology (Amsterdam, Netherlands), 122-123, 86-92 (2012-06-26)
It is well established that many pesticides used in the farming and horticultural industries are harmful to not only the target species they were developed for, but also other organisms. Organophosphates were introduced as a replacement for the organochlorines and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service