Skip to Content
Merck
All Photos(1)

Key Documents

45365

Supelco

Captafol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9Cl4NO2S
CAS Number:
Molecular Weight:
349.06
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC(Cl)C(Cl)(Cl)SN1C(=O)C2CC=CCC2C1=O

InChI

1S/C10H9Cl4NO2S/c11-9(12)10(13,14)18-15-7(16)5-3-1-2-4-6(5)8(15)17/h1-2,5-6,9H,3-4H2

InChI key

JHRWWRDRBPCWTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Caution

Restricted to professional users. Attention − Avoid exposure − obtain special instructions before use.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Futakuchi et al.
Cancer letters, 104(1), 37-41 (1996-06-24)
Effects of quinoline and captafol, both of which are hemangiocarcinogenic agents, were investigated in spontaneously hypertensive rats (SHR). Male SHR and Wistar Kyoto rats (WKY), the parent strain of SHR, were administered quinoline (0.2%) or captafol (0.15%) supplemented in the
M Rademaker
The Australasian journal of dermatology, 39(3), 164-167 (1998-09-16)
Forty-six farmers were patch tested to determine whether their dermatitis was secondary to an occupational allergen. Twenty-eight had a positive patch test of which 23 were thought relevant (definite or probable). In 20 of these cases, the allergen(s) was considered
H Tsuda et al.
Japanese journal of cancer research : Gann, 84(3), 230-236 (1993-03-01)
In a development trial for an initiation bioassay system, 7 known carcinogens and 1 suspected carcinogen were examined. In experiment 1, group 1 animals were initially subjected to partial hepatectomy (PH) 12 h before administration of diethylnitrosamine, 2-amino-3-methylimidazo[4,5-f]-quinoline (IQ), captafol
Final report on carcinogens background document for captafol.
Report on carcinogens background document for [substance name], (8-5974)(8-5974), i-xvi (2008-06-01)
F Ghillebaert et al.
Ecotoxicology and environmental safety, 32(1), 19-28 (1995-10-01)
Early life-stage survival, motility, and growth toxicity tests were carried out on common carp (Cyprinus carpio L.) endotrophic embryo, with two reference molecules (captafol and DNOC), from fertilization to the end of the first two-thirds period of mortalities by starvation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service