Skip to Content
Merck
All Photos(1)

Key Documents

37913

Supelco

Chlorfenapyr

PESTANAL®, analytical standard

Synonym(s):

4-Bromo-2-(4-chlorophenyl)-1-(ethoxymethyl)-5-(trifluoromethyl)-1H-pyrrol-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11BrClF3N2O
CAS Number:
Molecular Weight:
407.61
Beilstein:
6940152
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOCn1c(-c2ccc(Cl)cc2)c(C#N)c(Br)c1C(F)(F)F

InChI

1S/C15H11BrClF3N2O/c1-2-23-8-22-13(9-3-5-10(17)6-4-9)11(7-21)12(16)14(22)15(18,19)20/h3-6H,2,8H2,1H3

InChI key

CWFOCCVIPCEQCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Corine Ngufor et al.
Malaria journal, 16(1), 340-340 (2017-08-18)
Malaria control today is threatened by widespread insecticide resistance in vector populations. The World Health Organization (WHO) recommends the use of a mixture of unrelated insecticides for indoor residual spraying (IRS) and long-lasting insecticidal nets (LNs) or as a combination
Kimihiko Goto et al.
Journal of pesticide science, 44(4), 255-263 (2019-11-30)
Pyripyropene A (PP-A), a secondary metabolite produced by filamentous fungi, shows insecticidal activity against agricultural insect pests. Synthesized PP derivatives also show a narrow insecticidal spectrum but high insecticidal activities against such sucking pests. PP-A has a low eco-toxicological impact
James S Busse et al.
Journal of the science of food and agriculture, 99(5), 2579-2588 (2018-11-10)
Adverse air and soil temperatures are abiotic stresses that occur frequently and vary widely in duration and magnitude. Heat stress limits productivity of cool-weather crops such as potato (Solanum tuberosum) and may degrade crop quality. Stem-end chip defect is a
Aaron R Ashbrook et al.
Journal of economic entomology, 110(3), 1195-1202 (2017-04-12)
Insecticide resistance is a major impediment for effective control of Cimex lectularius L. Previous resistance detection studies with bed bugs have focused on certain pyrethroid, neonicotinoid, organochlorine, organophosphate, and carbamate insecticides. Within the pyrethroid class, resistance studies have mostly been
Richard M Oxborough et al.
Malaria journal, 14, 124-124 (2015-04-17)
The rapid selection of pyrethroid resistance throughout sub-Saharan Africa is a serious threat to malaria vector control. Chlorfenapyr is a pyrrole insecticide which shows no cross resistance to insecticide classes normally used for vector control and is effective on mosquito

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service