Skip to Content
Merck
All Photos(1)

Key Documents

34136

Supelco

HT-2 Toxin solution

100 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O8
CAS Number:
Molecular Weight:
424.48
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CC(C)CC(=O)O[C@H]1C[C@@]2(COC(C)=O)[C@H](O[C@@H]3[C@H](O)[C@@H](O)C2(C)C34CO4)C=C1C

InChI

1S/C22H32O8/c1-11(2)6-16(24)29-14-8-21(9-27-13(4)23)15(7-12(14)3)30-19-17(25)18(26)20(21,5)22(19)10-28-22/h7,11,14-15,17-19,25-26H,6,8-10H2,1-5H3/t14-,15+,17+,18+,19+,20+,21+,22+/m0/s1

InChI key

PNKLMTPXERFKEN-ZIOSACBISA-N

General description

Certan Vial

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Monaci et al.
Journal of chromatography. A, 1218(48), 8646-8654 (2011-10-29)
A sensitive and accurate method employing a single stage high resolution mass spectrometer equipped with a high-energy collision-dissociation cell (HCD) for the simultaneous determination of deoxynivalenol (DON), T-2 toxin (T-2) and HT-2 toxin (HT-2) in a processed bread model food
Vincenzo Lippolis et al.
Analytical and bioanalytical chemistry, 401(8), 2561-2571 (2011-09-09)
A rapid fluorescence polarization (FP) immunoassay has been developed for the simultaneous determination of T-2 and HT-2 toxins in naturally contaminated wheat samples. Syntheses of four fluorescein-labelled T-2 or HT-2 toxin tracers were carried out and their binding response with
Lian Li et al.
Animals : an open access journal from MDPI, 10(1) (2020-01-17)
Zearalenone (ZEA) and T-2 are the most common mycotoxins in grains and can enter the animal and human food-chain and cause many health disorders. To elucidate the toxic response profile, we stimulated bovine granulosa cells (GCs) with β-zearalenol or HT-2.
S De Baere et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(24), 2403-2415 (2011-07-20)
A sensitive and specific method for the quantitative determination of deoxynivalenol (DON), deepoxy-deoxynivalenol (DOM-1), T-2 toxin (T-2) and HT-2 toxin (HT-2) in animal body fluids (plasma and bile) using liquid chromatography combined with electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS) is
Francisco M Valle-Algarra et al.
Talanta, 84(4), 1112-1117 (2011-05-03)
There is a need to develop sensitive and accurate analytical methods for determining deoxynivalenol (DON), HT-2 toxin and T-2 toxin in paprika to properly assess the relevant risk of human exposure. An optimized analytical method for determination of HT-2 toxin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service