Skip to Content
Merck
All Photos(1)

Documents

33979

Supelco

Robenidine hydrochloride

VETRANAL®, analytical standard

Synonym(s):

1,3-Bis[(4-chlorobenzylidene)amino]guanidine monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13Cl2N5 · HCl
CAS Number:
Molecular Weight:
370.66
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].Clc1ccc(cc1)\C=N\NC(=N)N\N=C\c2ccc(Cl)cc2

InChI

1S/C15H13Cl2N5.ClH/c16-13-5-1-11(2-6-13)9-19-21-15(18)22-20-10-12-3-7-14(17)8-4-12;/h1-10H,(H3,18,21,22);1H/b19-9+,20-10+;

InChI key

LTWIBTYLSRDGHP-HCURTGQUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Robenidine hydrochloride is a coccidiostat and antibiotic, used in controlling coccidiosis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Robenidine hydrochloride has been used as a reference standard in the determination of robenidine hydrochloride in eggs using online solid-phase extraction with liquid chromatography coupled with tandem mass spectrometry (SPE-LC-MS/MS).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Heby et al.
Amino acids, 33(2), 359-366 (2007-07-05)
Trypanosomatids depend on spermidine for growth and survival. Consequently, enzymes involved in spermidine synthesis and utilization, i.e. arginase, ornithine decarboxylase (ODC), S-adenosylmethionine decarboxylase (AdoMetDC), spermidine synthase, trypanothione synthetase (TryS), and trypanothione reductase (TryR), are promising targets for drug development. The
Adnane Remmal et al.
Veterinary parasitology, 182(2-4), 121-126 (2011-07-06)
This study aims to assess the ability of essential oils (EOs) to destroy Eimeria oocyst in vitro using microscopic counting and 273 nm absorbing material release. A screening for the ability of ten EOs to destroy Eimeria oocyst was carried
Binh Nguyen et al.
Biophysical journal, 86(2), 1028-1041 (2004-01-30)
Many dicationic amidine compounds bind in the DNA minor groove and have excellent biological activity against a range of infectious diseases. Para-substituted aromatic diamidines such as furamidine, which is currently being tested against trypanosomiasis in humans, and berenil, which is
Multiclass analysis on repaglinide, flubendazole, robenidine hydrochloride and danofloxacin drugs
Al-Nahary TT, et al.
Arabian Journal of Chemistry, 6(1), 131-144 (2013)
B Stephen et al.
Veterinary parasitology, 69(1-2), 19-29 (1997-04-01)
Ten Eimeria field isolates from North Germany were studied in battery tests for sensitivity to selected anticoccidials. A high percentage of the Eimeria field isolates (9 out of 10) showed resistance to anticoccidials, mostly multiple resistance. Partial or complete resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service