Skip to Content
Merck
All Photos(3)

Documents

332488

Sigma-Aldrich

Crystal Violet lactone

Dye content 97 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29N3O2
CAS Number:
Molecular Weight:
415.53
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 97%

mp

180-183 °C (lit.)

λmax

354 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)C2(OC(=O)c3cc(ccc23)N(C)C)c4ccc(cc4)N(C)C

InChI

1S/C26H29N3O2/c1-27(2)20-11-7-18(8-12-20)26(19-9-13-21(14-10-19)28(3)4)24-16-15-22(29(5)6)17-23(24)25(30)31-26/h7-17H,1-6H3

InChI key

IPAJDLMMTVZVPP-UHFFFAOYSA-N

General description

Crystal Violet lactone (CVL) is a colorless lactone derivative of crystal violet. The CVL compound contains two dimethylaniline groups attached via a tetrahedral carbon atom to 6-dimethylaminophthalide at the third position.

Application

Crystal violet lactone is suitable for use in color-changing printing. It serves as a reversible color system with temperature change.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The percutaneous absorption of triarylmethane and phenoxazine type colour former components of carbonless copy papers.
B D Cameron et al.
The Annals of occupational hygiene, 30(1), 115-122 (1986-01-01)
Xiang Li et al.
The journal of physical chemistry. A, 115(16), 3746-3754 (2010-09-14)
Steady-state and picosecond time-resolved emission experiments are used to examine the excited-state charge transfer reaction of crystal violet lactone (CVL) in aprotic solvents. Solvatochromic analysis using a dielectric continuum model suggests dipole moments of 9-12 D for the initially excited
Kunihiro Ichimura et al.
Langmuir : the ACS journal of surfaces and colloids, 24(13), 6470-6479 (2008-06-10)
The solid phase adsorption of crystal violet lactone (CVL) on five types of Stober silica nanopowders with BET specific surface areas in the range of 50-800 m2/g under dry milling conditions was described for the first time. The hydrogen bonding
Uli Schmidhammer et al.
The journal of physical chemistry. A, 112(37), 8487-8496 (2008-08-22)
The intramolecular electron-transfer reaction in crystal violet lactone in polar aprotic solvents is studied with femtosecond transient absorption spectroscopy. The initially excited charge transfer state (1)CT A is rapidly converted into a highly polar charge transfer state (1)CT B. This
Jinlian H
Adaptive and Functional Polymers, Textiles and Their Applications (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service