Skip to Content
Merck
All Photos(2)

Key Documents

32673

Supelco

Ketoprofen-d3

VETRANAL®, analytical standard

Synonym(s):

2-(3-Benzoylphenyl)propionic acid-(methyl-d3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D3H11O3
CAS Number:
Molecular Weight:
257.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[2H]C([2H])([2H])C(C(O)=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/i1D3

InChI key

DKYWVDODHFEZIM-FIBGUPNXSA-N

Application

Ketoprofen-d3 has been used as a surrogate analytical standard for the determination of ketoprofen in sewage sludge samples by QuEChERS (quick, easy, cheap, effective, rugged and safe) extraction with on-line solid-phase extract purification and pre-concentration, followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS).
It may be used as an analytical standard for the determination of ketoprofen in compost from sewage sludge by ultrasound-assisted extraction (UAE) and salt-assisted liquid–liquid extraction (SALLE), followed by ultrahigh performance liquid chromatography (UHPLC) coupled to MS/MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaber Assaf et al.
Journal of pharmaceutical and biomedical analysis, 145, 414-422 (2017-07-22)
Investigations on the photochemical stability of pharmaceutical substances are mandatory in drug development and licensing as photo-induced degradation of an active pharmaceutical ingredient (API) may not only lead to decreased API concentrations but also to toxic or reactive products. Thus
Kristina Kotnik et al.
Chemosphere, 147, 114-123 (2016-01-15)
This study investigates the environmental fate of eight benzophenone derivatives (the pharmaceutical ketoprofen, its phototransformation products 3-ethylbenzophenone and 3-acetylbenzophenone, and five benzophenone-type UV filters) by evaluating their photolytic behaviour. In addition, the genotoxicity of these compounds and the produced photodegradation
Melissa M Melough et al.
Journal of agricultural and food chemistry, 65(24), 5049-5055 (2017-06-06)
Furocoumarins are a class of photoactive compounds found in several plant species and may be responsible for the observed association between consumption of citrus products and the risk of skin cancer. Furocoumarin contents of several foods have been reported previously
Innovative combination of QuEChERS extraction with on-line solid-phase extract purification and pre-concentration, followed by liquid chromatography-tandem mass spectrometry for the determination of non-steroidal anti-inflammatory drugs and their metabolites in sewage sludge.
Rossini D, et al.
Analytica Chimica Acta, 935, 269-281 (2016)
Sara Castiglioni et al.
Journal of hazardous materials, 384, 121441-121441 (2019-10-22)
In this work we evaluated the contamination of the water cycle in Como Bay by measuring 38 selected pharmaceuticals in two main wastewater treatment plant in Switzerland and in Italy, two influents (River Breggia and Cosia), lake water (epilimnion and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service