Skip to Content
Merck
All Photos(2)

Key Documents

32154

Supelco

Buparvaquone

VETRANAL®, analytical standard

Synonym(s):

2-[(4-tert-Butylcyclohexyl)methyl]-3-hydroxy-1,4-naphthoquinone, 2-[[4-(1,1-Dimethylethyl)cyclohexyl]methyl]-3-hydroxy-1,4-naphthalenedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)C1CCC(CC1)CC2=C(O)C(=O)c3ccccc3C2=O

InChI

1S/C21H26O3/c1-21(2,3)14-10-8-13(9-11-14)12-17-18(22)15-6-4-5-7-16(15)19(23)20(17)24/h4-7,13-14,24H,8-12H2,1-3H3

InChI key

KLLIVCPQDTYMLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bariş Saruhan et al.
Turkiye parazitolojii dergisi, 32(4), 317-321 (2009-01-22)
The aim of this study was to examine the efficacy of buparvaquone (Buparvon, ALKE, Istanbul) in the treatment of theileriosis in cattle. The causative agent T. annulata causes direct and indirect gross economical loss in Turkey. Theileriosis was microscopically diagnosed
Tracy Garnier et al.
The Journal of pharmacy and pharmacology, 59(1), 41-49 (2007-01-18)
As the part of a study to develop buparvaquone (BPQ) formulations for the treatment of cutaneous leishmaniasis, the topical delivery of BPQ and one of its prodrugs from a range of formulations was evaluated. In previous studies, BPQ and its
Gantala Venkatesh et al.
Drug development and industrial pharmacy, 36(6), 735-745 (2010-02-09)
The aim of this study was to prepare a lipid-based self-microemulsifying drug delivery system (SMEDDS) to increase the solubility and oral bioavailability of a poorly water-soluble compound, buparvaquone (BPQ). The solubility of BPQ was determined in various vehicles, and pseudo-ternary
Tracy Garnier et al.
The Journal of antimicrobial chemotherapy, 60(4), 802-810 (2007-08-24)
The efficacy of different formulations of the naphthoquinone buparvaquone and two phosphate prodrugs in in vivo models of both visceral and cutaneous leishmaniasis is described. Several topical formulations of buparvaquone containing acceptable excipients were tested in vivo against Leishmania major
Hassan Sharifiyazdi et al.
Veterinary parasitology, 187(3-4), 431-435 (2012-02-07)
Theileriosis is an economically important haemoprotozoal disease with high morbidity and mortality in cattle. Buparvaquone is very effective in the treatment of Theileria infections in cattle. The present study reported an outbreak of bovine tropical theileriosis in Fars Province, southern

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service