Skip to Content
Merck
All Photos(1)

Key Documents

1.00042

Supelco

Acetic anhydride

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Acetic anhydride, Acetyl acetate, Acetyl oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO)2O
CAS Number:
Molecular Weight:
102.09
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-564-8
NACRES:
NA.21
grade:
ACS reagent
form:
liquid

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

4 hPa ( 20 °C)

product line

EMSURE®
OmniSolv®

Assay

≥98.5% (GC)

form

liquid

autoignition temp.

330 °C

potency

630 mg/kg LD50, oral (Rat)
4320 mg/kg LD50, skin (Rabbit)

expl. lim.

2.0-10.2 % (v/v)

impurities

≤150 ppm Substances reducing potassium permanganate (as O)

evapn. residue

≤20 ppm

color

clearAPHA: ≤10

pH

3 (20 °C, 10 g/L in H2O)

mp

-73 °C

transition temp

flash point 49 °C

density

1.08 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤2 ppm
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤5 ppm

cation traces

Al: ≤0.5 ppm
B: ≤0.02 ppm
Ba: ≤0.1 ppm
Ca: ≤0.5 ppm
Cd: ≤0.05 ppm
Co: ≤0.02 ppm
Cr: ≤0.05 ppm
Cu: ≤0.02 ppm
Fe: ≤0.1 ppm
Mg: ≤0.1 ppm
Mn: ≤0.02 ppm
Ni: ≤0.02 ppm
Pb: ≤0.1 ppm
Sn: ≤0.1 ppm
Zn: ≤0.1 ppm
heavy metals (as Pb): ≤2 ppm

storage temp.

2-30°C

SMILES string

O(C(=O)C)C(=O)C

InChI

1S/C4H6O3/c1-3(5)7-4(2)6/h1-2H3

InChI key

WFDIJRYMOXRFFG-UHFFFAOYSA-N

Related Categories

General description

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Application

  • Insights into the Mechanism of Catalytic Activity of Plasmodium Parasite Malate-Quinone Oxidoreductase.: This study elucidates the catalytic mechanism of malate-quinone oxidoreductase in Plasmodium parasites, revealing potential targets for antimalarial drugs and insights into enzyme activity modulation by acetic anhydride (Ito et al., 2024).
  • Adsorption simulation of 2,4-D pesticide on novel zinc-based 2-amino-4-(1H-1,2,4-triazole-4-yl)benzoic acid coordination complexes using machine learning approach.: This research presents a novel adsorption method using zinc-based coordination complexes for the removal of pesticides, where acetic anhydride plays a crucial role in the synthesis of these complexes, demonstrating an innovative approach to environmental remediation (Mansab and Rafique, 2024).

Analysis Note

Purity (GC)≥ 98.5 %Identity (IR)conformsAppearanceclearColor≤ 10 HazenChloride (Cl)≤ 2 ppmSulfate (SO₄)≤ 5 ppmPhosphate (PO₄)≤ 5 ppmHeavy metals (as Pb)≤ 2 ppmReadily carbonizable substancesconformsSubstances reducing potassium permanganate (as O)≤ 150 ppmAl (Aluminium)≤ 0.5 ppmB (Boron)≤ 0.02 ppmBa (Barium)≤ 0.1 ppmCa (Calcium)≤ 0.5 ppmCd (Cadmium)≤ 0.05 ppmCo (Cobalt)≤ 0.02 ppmCr (Chromium)≤ 0.05 ppmCu (Copper)≤ 0.02 ppmFe (Iron)≤ 0.1 ppmMg (Magnesium)≤ 0.1 ppmMn (Manganese)≤ 0.02 ppmNi (Nickel)≤ 0.02 ppmPb (Lead)≤ 0.1 ppmSn (Tin)≤ 0.1 ppmZn (Zinc)≤ 0.1 ppmEvaporation residue
≤ 20 ppm≤ 20 ppm
Evaporation Number: 0.46

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service