Skip to Content
Merck
All Photos(1)

Key Documents

32001

Sigma-Aldrich

Sodium 5,5-diethylbarbiturate

puriss. p.a., ≥99.0% (NT)

Synonym(s):

5,5-Diethylbarbituric acid sodium salt, Barbitone, Sodium barbital, Veronal sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11N2NaO3
CAS Number:
Molecular Weight:
206.17
Beilstein:
3921202
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (NT)
solubility:
H2O: 0.1 g/mL, clear, colorless

grade

puriss. p.a.

Quality Level

Assay

≥99.0% (NT)

drug control

USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤30 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤500 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].CCC1(CC)C(=O)NC([O-])=NC1=O

InChI

1S/C8H12N2O3.Na/c1-3-8(4-2)5(11)9-7(13)10-6(8)12;/h3-4H2,1-2H3,(H2,9,10,11,12,13);/q;+1/p-1

InChI key

RGHFKWPGWBFQLN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 5,5-diethylbarbiturate (sodium barbital) is the sodium salt of 5,5-diethylbarbituric acid. Its crystal structure has been investigated by X-ray diffraction studies. Cell parameters have been reported as, a = 6.724 (1), b = 11.950 (2), c =12.129 (2)Å, Z = 4, Dm = 1.408g/cm-3.

Application

Sodium 5,5-diethylbarbiturate may be used to compose barbital buffer (pH 7.4), Michaelis buffer (pH 7.4) and veronal-buffered saline.

Biochem/physiol Actions

Sedative/hypnotic

Other Notes

Sales restrictions may apply

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 2 - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal structure of sodium 5, 5-diethylbarbiturate (sodium barbital).
Berking B and Craven BM.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 27(6), 1107-1115 (1971)
The properdin system and immunity: III. The zymosan assay of properdin.
Pillemer L, et al.
The Journal of Experimental Medicine, 103(1), 1-1 (1956)
Y Li et al.
Journal of thrombosis and haemostasis : JTH, 6(7), 1135-1143 (2008-04-25)
The adhesion ligand von Willebrand factor (VWF) is a multimeric glycoprotein that mediates platelet adhesion to exposed subendothelium. On endothelial cells, freshly released ultra-large (UL) VWF multimers form long string-like structures to which platelets adhere. The formation and elongation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service