Skip to Content
Merck
All Photos(1)

Key Documents

31706

Supelco

Nitrofurazone solution

VETRANAL®, 100 μg/mL in acetonitrile, analytical standard

Synonym(s):

Nitrofural solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O4
CAS Number:
Molecular Weight:
198.14
Beilstein:
1347298
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

NC(=O)N\N=C\c1ccc(o1)[N+]([O-])=O

InChI

1S/C6H6N4O4/c7-6(11)9-8-3-4-1-2-5(14-4)10(12)13/h1-3H,(H3,7,9,11)/b8-3+

InChI key

IAIWVQXQOWNYOU-FPYGCLRLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James R Johnson et al.
Antimicrobial agents and chemotherapy, 56(9), 4969-4972 (2012-07-04)
Two marketed antimicrobial-coated Foley catheters were compared for in vitro diffusible and contact-dependent inhibition of 11 urinary tract infection-associated microorganisms in an adherence-biofilm assay. Nitrofurazone-coated catheters significantly outperformed silver alloy-coated catheters for inhibitory activity, according to both inoculum broth and
Assessing catheter-associated urinary tract infection - Authors' reply.
Robert Pickard et al.
Lancet (London, England), 381(9877), 1536-1536 (2013-05-07)
Arnaud Gutierrez et al.
Nucleic acids research, 39(10), 4192-4201 (2011-02-09)
Escherichia coli SOS functions constitute a multifaceted response to DNA damage. We undertook to study the role of yafP, a SOS gene with unknown function. yafP is part of an operon also containing the dinB gene coding for DNA Polymerase
Man Chin Chung et al.
Current pharmaceutical design, 17(32), 3515-3526 (2011-11-15)
In therapeutics research, the nitro compounds are part of an important group of drugs with multiple pharmacological activities. However, in drug design, the inclusion of a nitro group in a molecule changes the physico-chemical and electronic properties and is associated
Fengyu Deng et al.
Molecular bioSystems, 8(5), 1446-1451 (2012-02-18)
The interaction of nitrofurazone (NF) and human serum albumin (HSA) has been studied by fluorescence spectroscopy, FT-IR spectroscopy and molecular modeling methods. The results showed that the fluorescence of HSA was quenched by NF in a static quenching mechanism. Thermodynamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service