Skip to Content
Merck
All Photos(1)

Key Documents

P5619

Supelco

Protein Standard

analytical standard, 2 mg/vial BSA

Synonym(s):

Protein Calibration Standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

form

lyophilized powder

analyte chemical class(es)

amino acids, peptides, proteins

concentration

2 mg/vial BSA

technique(s)

gel permeation chromatography (GPC): suitable

application(s)

food and beverages

format

neat

Application

Protein standard may be used for the calibration of detectors like refractive index detector or light scattering detector used in the size exclusion chromatographic analysis of proteins to determine their molecular weights of proteins and stoichiometry of the protein complex.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Size-exclusion chromatography with on-line light-scattering, absorbance, and refractive index detectors for studying proteins and their interactions
Wen J, et al.
Analytical Biochemistry, 240(2)`, 155-166 (1996)
Drew Ryan Peterson et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 178, 51-59 (2015-09-25)
Telomerase expression has long been linked to promotion of tumor growth and cell proliferation in mammals. Interestingly, telomerase activity (TA) has been detected in skeletal muscle for a variety of fish species. Despite this being a unique feature in fish
Herbert Michlmayr et al.
Toxins, 9(2) (2017-02-18)
Zearalenone (ZEN) is an estrogenic mycotoxin occurring in Fusarium-infected cereals. Glucosylation is an important plant defense mechanism and generally reduces the acute toxicity of mycotoxins to humans and animals. Toxicological information about ZEN-glucosides is limited due to the unavailability of
Hyuck-Jin Kwon et al.
Journal of environmental monitoring : JEM, 12(11), 2138-2144 (2010-10-05)
Rapid monitoring of the spreads of porcine reproductive and respiratory syndrome virus (PRRSV) was attempted using samples collected from nasal swabs of pigs and air samplers within an experimental swine building. An optofluidic device containing liquid-core waveguides was used to
Ana C A X De-Oliveira et al.
PloS one, 10(1), e0117842-e0117842 (2015-01-31)
Mouse cytochrome P450 (CYP) 2A5 is induced by inflammatory conditions and infectious diseases that down-regulate the expression and activity of most other CYP isoforms. Enhanced oxidative stress and nuclear factor (erythroid 2-related factor) 2 (Nrf2) transcription factor activation have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service