Skip to Content
Merck
All Photos(1)

Key Documents

31571

Supelco

Diafenthiuron

PESTANAL®, analytical standard

Synonym(s):

1-tert-Butyl-3-(2,6-diisopropyl-4-phenoxyphenyl)-thiourea, N-[2,6-Bis(1-methylethyl)-4-phenoxyphenyl]-N′-(1,1-dimethylethyl)-thiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N2OS
CAS Number:
Molecular Weight:
384.58
Beilstein:
8343025
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)c1cc(Oc2ccccc2)cc(C(C)C)c1NC(=S)NC(C)(C)C

InChI

1S/C23H32N2OS/c1-15(2)19-13-18(26-17-11-9-8-10-12-17)14-20(16(3)4)21(19)24-22(27)25-23(5,6)7/h8-16H,1-7H3,(H2,24,25,27)

InChI key

WOWBFOBYOAGEEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diafenthiuron is classified under the thiourea family of insecticides which are widely used for crop protection.

Application

Diafenthiuron may be used as a reference standard for the analysis of diafenthiuron in:
  • Cotton and groundnut oil by quick, easy, cheap, effective, rugged, and safe (QuEChERS) extraction procedure, low-temperature freezing and dispersive clean-up followed by quantification using gas chromatography (GC) equipped with electron capture detector (ECD) as well as flame photometric detector (FPD) and liquid chromatography-tandem mass spectrometry (LC-MS/MS).
  • Tomatoes by QuEChERS extraction and LC combined with triple quadrupole MS/MS with electrospray ionization source (ESI).
  • Fruit juice samples by ionic liquid-assisted liquid-phase microextraction based on the solidification of floating organic droplets (ILSFOD-LLME) and high performance liquid chromatography (HPLC) equipped with a variable-wavelength detector (VWD).
  • Water and wastewater by solid phase extraction (SPE) and LC combined with time-of-flight (TOF) MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

>300.2 °F - closed cup

Flash Point(C)

> 149 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

H Kayser et al.
Pest management science, 57(10), 975-980 (2001-11-07)
The thiourea insecticide/acaricide diafenthiuron represents a biologically inactive propesticide that requires transformation into the active carbodiimide derivative. The carbodiimide inhibits mitochondrial respiration by selective and covalent binding to the proteolipid (8 kDa) of Fo-ATPase in the inner membrane and to
Dissipation of foliar residues of diafenthiuron and its metabolites.
Y S Keum et al.
Bulletin of environmental contamination and toxicology, 68(6), 845-851 (2002-05-16)
The thiourea insecticide diafenthiuron inhibits mitochondrial ATPase in vitro and in vivo by its carbodiimide product.
F Ruder et al.
Biochemical Society transactions, 22(1), 241-244 (1994-02-01)
Young-Soo Keum et al.
Pest management science, 58(5), 496-502 (2002-05-10)
Diafenthiuron, 1-tert-butyl-3-(2,6-di-isopropyl-4-phenoxyphenyl)thiourea, is an effective insecticide and acaricide. Sunlight degradation of diafenthiuron in various aqueous solutions and pure hexane yielded two major identified products: 1-tert-butyl-3-(2,6-di-isopropyl-4-phenoxyphenyl)-carbodiimide (CGA-140,408) and 1-tert-butyl-3-(2,6-di-isopropyl-4-phenoxy-phenyl)urea (CGA-177,960). CGA-140,408 was further photo-transformed into CGA-177,960 by sunlight. Direct photolysis appeared
Multi-residue method for the determination of over 400 priority and emerging pollutants in water and wastewater by solid-phase extraction and liquid chromatography-time-of-flight mass spectrometry
Robles-Molina J, et al.
Journal of Chromatography A, 1350(3), 30-43 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service