Skip to Content
Merck
All Photos(3)

Documents

27086

Supelco

Chromoionophore I

Selectophore

Synonym(s):

3-Octadecanoylimino-7-(diethylamino)-1,2-benzophenoxazine, 9-(Diethylamino)-5-(octadecanoylimino)-5H-benzo[a]phenoxazine, N-Octadecanoyl-Nile blue, ETH 5294

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H53N3O2
CAS Number:
Molecular Weight:
583.85
Beilstein:
3576427
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

solid

mp

91-93 °C (lit.)

fluorescence

λex 614 nm; λem 663 nm (protonated)

SMILES string

CCCCCCCCCCCCCCCCCC(=O)\N=C1/C=C2Oc3cc(ccc3N=C2c4ccccc14)N(CC)CC

InChI

1S/C38H53N3O2/c1-4-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25-37(42)39-34-29-36-38(32-24-22-21-23-31(32)34)40-33-27-26-30(28-35(33)43-36)41(5-2)6-3/h21-24,26-29H,4-20,25H2,1-3H3/b39-34+

InChI key

MYWBZQJIXVRCJC-JQJRJOTLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromoionophore I (ETH 5294, 3-octadecanoylimino-7-diethylamino-l,2-benzophenoxazine) is a H+-selective neutral chromoionophore, which is lipophilic in nature and is a highly basic dye molecule.
Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Chromoionophore I may be used for the test zone preparation in the multianalyte sensor, during environmental monitoring, using photographic digital camera.
Assay of K+ and of Ca2+ activity with solvent polymeric optode membrane based on ETH 5294 and potassium ionophore I (valinomycin)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmental monitoring using a conventional photographic digital camera for multianalyte disposable optical sensors.
Lapresta FA and Vallvey C.
Analytica Chimica Acta, 706 (2), 328-337 (2011)
Anna Baranowska-Korczyc et al.
The Analyst, 145(16), 5594-5602 (2020-08-11)
Core-shell modified nanofiber mats were used as ion-selective membranes for the first time. Keeping the overall macroscopic size of the sensing element the same as for classical plasticized poly(vinyl chloride) membranes, herein the proposed nanofiber based systems resulted in ultrathin
L Lvova et al.
Analytica chimica acta, 988, 96-103 (2017-09-17)
A systematic study of a series of diaza-18-crown-6 8-hydroxyquinoline (DCHQ) chemosensors, devoted to Mg(II) ion detection, was performed. Functionalization of DCHQ by peripheral substituents allowed the development of novel all-solid-state optodes via inclusion inside PVC-based polymeric films. The influence on
Fatehy M Abdel-Haleem et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 201, 98-104 (2018-05-08)
Preparation of novel salicylate-selective optical sensors (bulk optodes) was performed and applied successfully for salicylate determination in pharmaceutical formulations, Aspirin® and Aspocid®. t-butyl calix[4]arene ionophore was incorporated in a plasticized poly (vinyl-chloride) membrane containing the chromoionophore ETH5294 (O1) or ETH7075
Design and characterization of a novel ammonium ion selective optical sensor based on neutral ionophores.
Seiler K
Analytical Sciences, 5 (5), 557-561 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service