
Chromoionophore I

Synonym(s):
3-Octadecanoylimino-7-(diethylamino)-1,2-benzophenoxazine, 9-(Diethylamino)-5-(octadecanoylimino)-5H-benzo[a]phenoxazine, N-Octadecanoyl-Nile blue, ETH 5294
Empirical Formula (Hill Notation):
C38H53N3O2
CAS Number:
Molecular Weight:
583.85
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing