Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,5-Diphenylcarbazide
Synonym(s):
1,5-Diphenylcarbazide, 1,5-Diphenylcarbohydrazide, sym.-Diphenylcarbazide
Linear Formula:
C
6
H
5
NHNHCONHNHC
6
H
5
CAS Number:
140-22-7
Molecular Weight:
242.28
EC Number:
205-403-7
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (2)
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.03091
1,5-Diphenylcarbazide,
GR for analysis and redox indicator ACS,Reag. Ph Eur
Expand
View Pricing
259225
1,5-Diphenylcarbazide,
ACS reagent
Expand
View Pricing
D7766
1,5-Diphenylcarbazide,
reagent grade
Expand
View Pricing
33152
1,5-Diphenylcarbazide,
reag. Ph. Eur., ≥98.0%
Expand
View Pricing