Skip to Content
Merck
All Photos(2)

Key Documents

242969

Sigma-Aldrich

Sodium diphenylamine-4-sulfonate

ACS reagent

Synonym(s):

4-(Phenylamino)benzenesulfonic acid sodium salt, Diphenylamine-4-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10NNaO3S
CAS Number:
Molecular Weight:
271.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent

grade

ACS reagent

Quality Level

technique(s)

titration: suitable

SMILES string

[Na+].[O-]S(=O)(=O)c1ccc(Nc2ccccc2)cc1

InChI

1S/C12H11NO3S.Na/c14-17(15,16)12-8-6-11(7-9-12)13-10-4-2-1-3-5-10;/h1-9,13H,(H,14,15,16);/q;+1/p-1

InChI key

DGXTZMPQSMIFEC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium diphenylamine-4-sulfonate has the ability to form colored complexes with metal ions. Hence it is commonly used as an indicator to estimate the concentration of metal ions. It is also an efficient quencher of electrochemiluminescence of tris (2, 2′-bipyridine)-ruthenium (II).

Application

Sodium diphenylamine-4-sulfonate may be used in the synthesis of poly-(aniline-co-N-(4-sulfophenyl)aniline via polymerization with aniline. It may also be used as a reductant in the synthesis of branched gold nanocrystals from gold(III) chloride trihydrate in the presence of polyethylene glycol. These nanocrystals have potential application as electrocatalysts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction products between sodium diphenyl-amine-4-sulfonate and hydrated LaCl3.
Machado LC, et al.
Journal of Thermal Analysis and Calorimetry, 75(2), 615-621 (2004)
Quenching of the Electrochemiluminescence of Tris (2, 2'-bipyridine)-ruthenium (II) by Sodium Diphenylamine-4-sulfonate
Zhu Y, et al
Analytical Sciences, 25(6), 785-788 (2009)
The effects of several sulfonates on thermal and fire retardant properties of poly (methyl methacrylate) and polystyrene.
He J, et al.
Polymers For Advanced Technologies, 25(2), 160-167 (2014)
High-yield Synthesis of Branched Gold Nanocrystals by a Sodium Diphenylamine-4-Sulfonate Reduction Process in Polyethylene Glycol Aqueous Solution.
Liu Q, et al.
Chin. J. Chem., 28(4), 537-542 (2010)
Synthesis and properties of novel water-soluble conducting polyaniline copolymers.
Nguyen MT, et al.
Macromolecules, 27(13), 3625-3631 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service