Skip to Content
Merck
All Photos(1)

Key Documents

27096

Sigma-Aldrich

Chromium(III) chloride hexahydrate

purum p.a., ≥98.0% (RT)

Synonym(s):

Chromium trichloride hexahydrate, Hexaaquachromium(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrCl3 · 6H2O
CAS Number:
Molecular Weight:
266.45
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

purum p.a.

Quality Level

Assay

≥98.0% (RT)

form

solid

reaction suitability

core: chromium
reagent type: catalyst

density

1.76 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤300 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤100 mg/kg
K: ≤100 mg/kg
Na: ≤300 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Cr](Cl)Cl

InChI

1S/3ClH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

LJAOOBNHPFKCDR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Chromium(III) chloride (CrCl3. 6H2O) hexahydrate can be used:
  • As a source of chromium(II), which is in turn used as a catalyst in iodovinylation of aldehydes (Takai reaction).
  • In the synthesis of chromium(III) complexes of tetradentate Schiff bases.
  • In the preparation of Cr(III)-fatty acid complexes which can further be used as a catalyst for the synthesis of 1-monoacyIgIyceroIs.
  • In combination with choline chloride as a deep eutectic solvent, which can be utilized as a catalyst for the esterification of formic and acetic acid with alcohols.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization, and spectroscopic studies of tetradentate Schiff base chromium (III) complexes
Aranha PE, et al.
Polyhedron, 26(7), 1373-1382 (2007)
Deep eutectic solvent choline chloride 2CrCl3.6H2O: an efficient catalyst for esterification of formic and acetic acid at room temperature
Cao J, et al.
Royal Society of Chemistry Advances, 6(26), 21612-21616 (2016)
An alternative procedure in the Takai reaction using chromium (III) chloride hexahydrate as a convenient source of chromium (II)
Auge J, et al.
Synthetic Communications, 33(21), 3733-3739 (2003)
Preparation of 1-monoacyIgIyceroIs from gIycidoI and fatty acids cataIyzed by the chromium (III)-fatty acid system
Janivs R, et al.
European Journal of Lipid Science and Technology, 102(5), 351-354 (2000)
Igor Shuryak et al.
Scientific reports, 9(1), 11361-11361 (2019-08-08)
Exposure to chronic ionizing radiation (CIR) from nuclear power plant accidents, acts of terrorism, and space exploration poses serious threats to humans. Fungi are a group of highly radiation-resistant eukaryotes, and an understanding of fungal CIR resistance mechanisms holds the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service