Skip to Content
Merck
All Photos(1)

Key Documents

21310

Sigma-Aldrich

(±)-Camphor

purum, synthetic, ≥95.0% (GC)

Synonym(s):

1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1907611
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.2 (vs air)

Quality Level

vapor pressure

4 mmHg ( 70 °C)

grade

purum

Assay

≥95.0% (GC)

quality

synthetic

expl. lim.

3.5 %

impurities

≤0.5% water

bp

204 °C (lit.)

mp

175-177 °C (lit.)

solubility

ethanol: soluble 1 g/10 mL, clear, colorless
water: soluble

functional group

ketone

SMILES string

[H][C@](CC1=O)(CC2)C(C)(C)[C@]12C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly diastereoselective synthesis of new optically active aminoalcohols in one step from (+)-camphor and (-)-fenchone.
Genov M, et al.
Tetrahedron Asymmetry, 8(11), 1869-1876 (1997)
Planer nano-graphenes from camphor by CVD.
Somani PR, et al.
Chemical Physics Letters, 430(1), 56-59 (2006)
Dong-Bin Dang et al.
Chemical communications (Cambridge, England), 49(22), 2243-2245 (2013-02-12)
A series of 3D homochiral manganese-lanthanide frameworks have been synthesized based on chiral camphoric acid. In the heterometallic features, D-camphoric acids are unprecedentedly embedded in three coordination modes.
Hiroyuki Yamashita et al.
Pharmaceutical research, 30(1), 70-80 (2012-08-22)
Although a number of studies have reported that cocrystals can form by heating a physical mixture of two components, details surrounding heat-induced cocrystal formation remain unclear. Here, we attempted to clarify the thermal behavior of a physical mixture and cocrystal
Christian Merten et al.
The journal of physical chemistry. A, 116(27), 7329-7336 (2012-06-06)
In the present work, the first observation of strong resonance Raman optical activity (RROA) involving more that one resonant electronic state is reported. The chiral transition metal complex bis-(trifluoroacetylcamphorato) copper(II), abbreviated Cu(tfc)(2), exhibits both resonance Raman (RR) and RROA spectra

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service