Skip to Content
Merck
All Photos(1)

Key Documents

18106

Sigma-Aldrich

Sodium benzoate

puriss., meets analytical specification of Ph. Eur., BP, FCC, E211, 99.0-100.5% (calc. to the dried substance), powder

Synonym(s):

BENZOTRON, Benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39024301
PubChem Substance ID:
NACRES:
NA.21
form:
powder
solubility:
water: soluble 556 g/L

grade

puriss.

Quality Level

Assay

99.0-100.5% (calc. to the dried substance)

form

powder

quality

meets analytical specification of Ph. Eur., BP, FCC, E211

impurities

acidity or alkalinity, complies
oxidizable impurities, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.03% halogenated compounds (as Cl)

loss

≤1% loss on drying, 105 °C

mp

>300 °C (lit.)

solubility

water: soluble 556 g/L

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Cu: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤2 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

functional group

phenyl

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium benzoate can be used as areactant to synthesize:
  • 2-Phenoxyethyl benzoate via phase-transfercatalyzed esterification with 2-phenoxyethyl bromide.
  • Ynones by Pd free coupling reactionwith terminal alkynes in the presence of cyanuric chloride and magnesiumchloride.
It can also be used as a catalyst to synthesize:
  • 3-Methyl-4-arylmethylene-isoxazol-5(4H)-onesby reacting ethyl acetoacetate, hydroxylamine hydrochloride, and aromaticaldehydes.
  • 4-Pyrazolylmethylene-3-phenylisoxazol-5(4H)-onesvia three-component cyclocondensation of pyrazole-4-carbaxaldehyde withβ-ketoesters and hydroxylamine hydrochloride.

Legal Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 121-121 (1994)
Huey-Jen Tsay et al.
Neurotoxicology and teratology, 29(5), 562-569 (2007-07-24)
Sodium benzoate (SB) is a commonly used food preservative and anti-microbial agent in many foods from soup to cereals. However, little is known about the SB-induced toxicity and teratogenicity during early embryonic development. Here, we used zebrafish as a model
Saurav Brahmachari et al.
Journal of immunology (Baltimore, Md. : 1950), 179(1), 275-283 (2007-06-21)
Experimental allergic encephalomyelitis (EAE) is the animal model for multiple sclerosis. This study explores a novel use of sodium benzoate (NaB), a commonly used food additive and a Food and Drug Administration-approved nontoxic drug for urea cycle disorders, in treating
H M Pylypiw et al.
Journal of chromatography. A, 883(1-2), 299-304 (2000-07-26)
A rapid and reliable method is presented for the determination of the preservatives sodium benzoate and potassium sorbate in fruit juices, sodas, soy sauce, ketchup, peanut butter, cream cheese, and other foods. The procedure utilizes high-performance liquid chromatography (HPLC) followed
Jia Li et al.
Environmental science and pollution research international, 18(9), 1567-1573 (2011-05-18)
This study investigated the effect of copper on chemical oxygen demand (COD) removal efficiency and on the properties (mainly settling and dewatering) and the composition of extracellular polymeric substances (EPS) when 20 mg/L Cu(II) was continuously dosed to a sequencing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service