Skip to Content
Merck
All Photos(2)

Documents

13180

Supelco

Benzylamine

for GC derivatization, LiChropur, ≥99.0%

Synonym(s):

α-Aminotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NH2
CAS Number:
Molecular Weight:
107.15
Beilstein:
741984
EC Number:
MDL number:
UNSPSC Code:
12000000
eCl@ss:
39030404
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥99.0% (GC)
≥99.0%

form

liquid

quality

LiChropur

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.543 (lit.)
n20/D 1.543

bp

184-185 °C (lit.)

mp

10 °C (lit.)

solubility

H2O: soluble

density

0.981 g/mL at 25 °C (lit.)

SMILES string

NCc1ccccc1

InChI

1S/C7H9N/c8-6-7-4-2-1-3-5-7/h1-5H,6,8H2

InChI key

WGQKYBSKWIADBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzylamine, a primary amine, can be used as a fluorescence derivatization agent for the sensitive and selective determination of analytes by chromatography techniques coupled with fluorescence detection.

Application

Benzylamine may be used as a derivatization agent to increase the sensitivity of 5-hydroxyindoles, catecholamines and catechols in biological samples prior to their determination using high performance liquid chromatography (HPLC) coupled with fluorescence detection.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous determination of norepinephrine, serotonin, and 5-hydroxyindole-3-acetic acid in microdialysis samples from rat brain by microbore column liquid chromatography with fluorescence detection following derivatization with benzylamine
Yoshitake T, et al.
Analytical Biochemistry, 312(2), 125-133 (2003)
The Derivatization of Proquinoidal Analytes with 1, 2-Diphenylethane-1, 2-Diamine (DPE) and Benzylamine (BA): An Investigation of Products, Yields, Kinetics and Reagent Selectivity
Woods JM, et al.
Chromatographia, 78(3-4), 163-178 (2015)
Kaoru Fujino et al.
Journal of chromatography. A, 1012(2), 169-177 (2003-10-03)
A highly selective and sensitive method for the simultaneous determination of 5-hydroxyindoles and catechols (serotonin, norepinephrine, dopamine and related compounds) by high-performance liquid chromatography with fluorescence detection is described. The method is based on the two-step precolumn derivatization of 5-hydroxyindoles
Danielle M Shacklady-McAtee et al.
Organic letters, 13(13), 3490-3493 (2011-06-08)
A nickel(0) catalyst effectively mediates the cyclization of N-benzoyl aminals in the presence of a stoichiometric Lewis acid. This method enables preparation of a variety of isoindolinones with substitution on the benzoyl fragment and C-3 carbon. This reaction likely proceeds
Farid Oukacine et al.
Analytical chemistry, 84(7), 3302-3310 (2012-03-28)
A new methodology for an antibacterial assay based on capillary electrophoresis with multiple UV detection points has been proposed. The possible antibacterial activity of cationic molecules on bacteria (Gram-positive and Gram-negative) is studied by detecting the bacteria before, during, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service