Skip to Content
Merck
All Photos(2)

Key Documents

128945

Sigma-Aldrich

Phenethylamine

99%

Synonym(s):

2-Phenethylamine, β-Phenylethylamine, 2-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
507488
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.18 (vs air)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

197-200 °C (lit.)

density

0.962 g/mL at 20 °C (lit.)

functional group

amine
phenyl

SMILES string

NCCc1ccccc1

InChI

1S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6-7,9H2

InChI key

BHHGXPLMPWCGHP-UHFFFAOYSA-N

Gene Information

human ... AOC3(8639)
mouse ... Aoc3(11754)
rat ... Htr2a(29595)

Looking for similar products? Visit Product Comparison Guide

Application

Phenethylamine was used to study the mass spectra of [2-(4-iodo-2,5-dimethoxyphenyl)-N-[(2,3-methylenedioxyphenyl)methyl]ethanamine], a psychoactive substance.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Deepak Narang et al.
The Journal of pharmacology and experimental therapeutics, 351(1), 164-171 (2014-08-15)
The trace amine β-phenylethylamine (PEA) is normally present in the body at low nanomolar concentrations but can reach micromolar levels after ingestion of drugs that inhibit monoamine oxidase and primary amine oxidase. In vivo, PEA elicits a robust pressor response
Douwe de Boer et al.
Pharmacy world & science : PWS, 26(2), 110-113 (2004-04-17)
To collect data related to phenethylamine drugs-of-abuse of the 2C-series, to review possible health risks of their use and to discuss legal counter actions of authorities in the European Union (EU). Dutch smartshops. In the period of 1994-2002, all products
Hee-Jin Jeong et al.
Methods and protocols, 3(2) (2020-06-18)
The problem of illicit drug use and addiction is an escalating issue worldwide. As such, fast and precise detection methods are needed to help combat the problem. Herein, the synthesis method for an anti-methamphetamine Quenchbody (Q-body), a promising sensor for
Nickolaj J Petersen et al.
Electrophoresis, 33(6), 1021-1031 (2012-04-25)
The traditional way of calculating mobility and peak areas in capillary electrophoresis does not take into account the changes in the buffer viscosity at different thermostatic control and that the analytes may accelerate during the individual runs due to Joule
Karolina Sekuła et al.
Rapid communications in mass spectrometry : RCM, 27(18), 2081-2090 (2013-08-15)
In recent years, the phenomenon of uncontrolled distribution of new psychoactive substances that were marketed without prior toxicological studies has been observed. Because many designer drugs are related in chemical structure, the potential for misidentifying them is an important problem.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service