Skip to Content
Merck
All Photos(2)

Documents

04607

Supelco

Methyl eugenol

analytical standard

Synonym(s):

Eugenol methyl ether, 4-Allyl-1,2-dimethoxybenzene, Eugenyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C6H3(OCH3)2
CAS Number:
Molecular Weight:
178.23
Beilstein:
1910871
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

>98% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.534 (lit.)

bp

254-255 °C (lit.)

mp

−4 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(CC=C)cc1OC

InChI

1S/C11H14O2/c1-4-5-9-6-7-10(12-2)11(8-9)13-3/h4,6-8H,1,5H2,2-3H3

InChI key

ZYEMGPIYFIJGTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl eugenol is a phenyl methyl ether belonging to the class of alkylbenzenes. It is derived from eugenol and commonly identified as one of the important components of basil essential oil. It is widely used as an active pesticide ingredient and also employed as an insect parapheromone in insect traps, to attract certain species such as oriental fruit flies.

Application

Methyl eugenol may be used as an analytical reference standard for the quantification of the analyte in the following:        
  • Essential oil of Pimenta pseudocaryophyllus using high-performance liquid chromatography (HPLC). The purity of the extracted compound is then determined by gas chromatography coupled to flame ionization detector (GC/FID).       
  • Stem bark of Cinnamomum zeylanicum Blume using reversed-phase high-performance liquid chromatography (RP-HPLC) with UV-visible detection.       
  • Rosa hybrida using gas-chromatography coupled to mass spectrometry (GC-MS) technique.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: cannabis cinnamomum

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Muta. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benefits of the Mediterranean Diet in the Elderly Patient (2018)
The Western Herbal Tradition E-Book: 2000 years of medicinal plant knowledge (2010)
Isolation of chavibetol and methyleugenol from essential oil of Pimenta pseudocaryophyllus by high performance liquid chromatography
Niculau SdE, et al.
Molecules (Basel), 23(11), 2909-2909 (2018)
Simultaneous quantitative determination of cinnamaldehyde and methyl eugenol from stem bark of Cinnamomum zeylanicum Blume using RP-HPLC
Gursale A, et al.
Journal of Chromatographic Science, 48(1), 59-62 (2010)
Alexander T Cartus et al.
Toxicological sciences : an official journal of the Society of Toxicology, 129(1), 21-34 (2012-05-23)
Methyleugenol (1) is a constituent of many foods, in particular of herbal spices, and is used as flavoring agent in foodstuffs and as fragrance in cosmetics. 1 has been found to be carcinogenic in rodents, its metabolite, 1-hydroxymethyleugenol (2) acting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service