D136905
1,2-Dimethoxy-4-propenylbenzene
99%
Synonym(s):
Isoeugenyl methyl ether
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH=CHC6H3(OCH3)2
CAS Number:
Molecular Weight:
178.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
description
mixture of isomers
Quality Level
Assay
99%
form
liquid
refractive index
n20/D 1.568 (lit.)
bp
262-264 °C (lit.)
mp
98-100 °C (lit.)
density
1.05 g/mL at 25 °C (lit.)
SMILES string
COc1ccc(\C=C\C)cc1OC
InChI
1S/C11H14O2/c1-4-5-9-6-7-10(12-2)11(8-9)13-3/h4-8H,1-3H3/b5-4+
InChI key
NNWHUJCUHAELCL-SNAWJCMRSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gang Wei et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 29(8), 764-768 (2004-10-28)
To establish the method of fingerprint analysis on volatile oil in rhizome of Acorus tatarinowii by GC-MS, and to study the main characteristic components. The main components of 10 samples were determined by GC-MS. The injector temperature was 250 degrees
[In vitro study of the expectorant effect of phenylpropane derivatives of hazelwort. 12. Active substances form Asarum europaeum. L].
L Gracza
Planta medica, 42(2), 155-159 (1981-06-01)
Alexander T Cartus et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(9), 1727-1733 (2011-06-03)
Methylisoeugenol (1,2-dimethoxy-4-propenylbenzene, 1) is a minor constituent of essential oils, naturally occurring as a mixture of cis/trans isomers. 1 is a U.S. Food and Drug Administration-approved food additive and has been given "Generally Recognized as Safe" status. Previously, metabolism of
Paul-Georges Rossi et al.
Journal of agricultural and food chemistry, 55(18), 7332-7336 (2007-08-10)
The essential oil of wild Daucus carota L. obtained from aerial parts at the end of the flowering stage (DCEO) was reported as antimicrobial against the human enteropathogen Campylobacter jejuni. The aim of the present study was to extend this
A call for full ingredient labelling and uniform nomenclature on medicaments.
A E Pink et al.
The British journal of dermatology, 161(2), 489-490 (2009-06-24)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service