Skip to Content
Merck
All Photos(1)

Key Documents

04500

Sigma-Aldrich

4-Ethylmorpholine

≥97.0% (GC)

Synonym(s):

N-Ethylmorpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
Beilstein:
102969
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39162412
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

impurities

≤3% 4-methylmorpholine

refractive index

n20/D 1.441 (lit.)
n20/D 1.442

bp

139 °C (lit.)

mp

−63 °C (lit.)

density

0.91 g/mL at 20 °C (lit.)

functional group

ether

SMILES string

CCN1CCOCC1

InChI

1S/C6H13NO/c1-2-7-3-5-8-6-4-7/h2-6H2,1H3

InChI key

HVCNXQOWACZAFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aliaksandr Druz et al.
RNA biology, 10(2), 287-300 (2013-01-29)
MicroRNAs (miRNAs) have been found to be involved in cancer initiation, progression and metastasis and, as such, have been suggested as tools for cancer detection and therapy. In this work, a large-scale screening of the complete miRNA mimics library demonstrated
Andreas Zettl et al.
The American journal of pathology, 161(5), 1635-1645 (2002-11-05)
Genetic alterations in enteropathy-type T-cell lymphoma (ETL) are unknown so far. In this series, 38 cases of ETL were analyzed by comparative genomic hybridization (CGH). CGH revealed chromosomal imbalances in 87% of cases analyzed, with recurrent gains of genetic material
Muhammad Hamza Bokhari et al.
Communications biology, 4(1), 1108-1108 (2021-09-23)
The activation of thermogenesis in adipose tissue has emerged as an important target for the development of novel anti-obesity therapies. Using multi-well isothermal microcalorimetry, we have demonstrated that mature murine brown and brite adipocytes produce quantifiable heat upon β3-AR stimulation
Yun Wang et al.
Molecules (Basel, Switzerland), 26(18) (2021-09-29)
Mn and Fe BPMEN complexes showed excellent reactivity in catalytic oxidation with an excess of co-reagent (CH3COOH). In the straight line of a cleaner catalytic system, volatile acetic acid was replaced by SiO2 (nano)particles with two different sizes to which
D R Twardzik et al.
Proceedings of the National Academy of Sciences of the United States of America, 69(1), 274-277 (1972-01-01)
Cell suspensions derived from a mouse plasmacytoma (RPC-20) that secretes an immunoglobulin light chain containing N-terminal pyroglutamic acid can synthesize protein in vitro. Chromatographic examination of an enzymatic digest of protein labeled with glutamic acid shows only labeled glutamic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service