Skip to Content
Merck
All Photos(2)

Key Documents

04058

Sigma-Aldrich

1-Propanesulfonic acid

technical, ≥99.0% (T)

Synonym(s):

Propanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SO3H
CAS Number:
Molecular Weight:
124.16
Beilstein:
1747491
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical

Assay

≥99.0% (T)

form

liquid

color

clear brown

refractive index

n20/D 1.44

density

1.25 g/mL at 20 °C (lit.)

functional group

sulfonic acid

SMILES string

CCCS(O)(=O)=O

InChI

1S/C3H8O3S/c1-2-3-7(4,5)6/h2-3H2,1H3,(H,4,5,6)

InChI key

KCXFHTAICRTXLI-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I A Casalinuovo et al.
European review for medical and pharmacological sciences, 21(6), 1413-1420 (2017-04-08)
Candida albicans biofilm is frequently found on artificial surfaces and the infections related to biofilm are difficult to eliminate, as they require the removal of artificial devices and treatment with antifungal drugs. Nowadays, fungal growth in biofilms is difficult to
Songnian Liu et al.
Scientific reports, 10(1), 9023-9023 (2020-06-04)
Cardiovascular disease (CVD) is the major cause of morbidity, mortality, and health care costs in the United States, and possibly around the world. Among the various risk factors of CVD, environmental and dietary exposures to mercury (Hg), a highly toxic
Gayani S Jayasooriya et al.
Anesthesia and analgesia, 132(1), 231-239 (2020-08-29)
Nitroglycerin is used for acute reduction in uterine tone. Prolonged oxytocin exposure causes desensitization of oxytocin receptors. It is unknown if nitroglycerin exposure impacts the subsequent action of oxytocin in the setting of oxytocin receptor desensitization. This study investigated the
Zahra Salehi et al.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology, 37(10), 1841-1846 (2018-07-08)
With regard to increasing number of antifungal-resistant dermatophytes, antifungal susceptibility testing of dermatophytes serves as a useful tool in managing clinical dermatophytosis. This study aimed to determine antifungal susceptibility profile of clinically important dermatophytes and determination of point mutations in
Kathryn L Kay et al.
Frontiers in microbiology, 8, 1404-1404 (2017-08-12)
Shiga toxin producing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service