Skip to Content
Merck
All Photos(1)

Documents

02538

Millipore

Raka-Ray Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Lactic Acid Bacteria Selective Agar Base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

N-acetyl glucosamine, 0.5 g/L
agar, 17.0 g/L
betaine hydrochloride, 2.0 g/L
casein enzymic hydrolysate, 20.0 g/L
diammonium hydrogen-citrate, 2.0 g/L
fructose, 5.0 g/L
glucose, 5.0 g/L
liver concentrate, 1.0 g/L
magnesium sulphate, 2.0 g/L
maltose, 10.0 g/L
manganese sulfate, 0.66 g/L
monopotassium phosphate, 2.0 g/L
potassium aspartate, 2.5 g/L
potassium glutamate, 2.5 g/L
yeast extract, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

5.4±0.2 (25 °C)

application(s)

agriculture
bioburden testing
food and beverages

microbiology

shipped in

wet ice

storage temp.

2-8°C

suitability

Pediococcus spp.
selective for Lactobacillus spp.

General description

Raka-Ray Agar was formulated by Saha, Sondag and Middlekauff to monitor the brewing process and analyze it for a wide range of bacteria. The media is found to be superior to several other media for the cultivation of Lactobacilli and Pediococci. It also suppresses the growth of non-lactic acid facultative bacteria that are often associated with lactic beer spoilage.

Application

Raka-Ray Agar is recommended for selective isolation of lactic acid bacteria encountered in beer and the brewing process.

Preparation Note

Suspend 38.5 g in 500 ml of distilled water. Bring to the boil and dissolve the medium completely. Distribute into tubes or bottles and sterilize by autoclaving at 121°C for 15 minutes. Cool to 50-55°C and add contents of 1vial of lactic acid bacteria selective supplement (Cat. No. 14121). Add 3.5 ml of Cycloheximide solution (Cat. No. 18079). Mix well and pour into sterile petri dishes or dispense as desired.

Analysis Note

Casein enzymic hydrolysate, liver concentrate, yeast extract provide carbon, nitrogen, amino acids, minerals, vitamins, trace elements and other essential nutrients for growth. Potassium aspartate and potassium glutamate are additional sources of amino acids. Maltose is to detect lactobacilli, which cannot utilize glucose as a carbon source and fructose is the carbon source of Lactobacillus fructivorans. Glucose is needed as the fermentative energy source for the Pediococci. Sorbitan mono-oleate act as a stimulant for lactic acid bacteria in general. Also, N-acetyl glucosamine, liver concentrate yeast extract and betaine hydrochloride are used as growth stimulating agents. Diammonium hydrogen-citrate and Monopotassium phosphate are the buffering agents while Magnesium and Manganese are important trace elements for Lactobacillaceae. The addition of phenylethanol inhibits Gram-negative organisms, while amphotericin B and cycloheximide inhibits growth of yeasts and molds.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sixty years of Amphotericin B: An Overview of the Main Antifungal Agent Used to Treat Invasive Fungal Infections
Infectious diseases and therapy, 1-33 (2021)
European Brewing Convention
Journal of the Institute of Brewing, 87, 303-303 (1981)
C. Van Keer et al.
Journal of the Institute of Brewing, 89, 361-361 (1983)
The genera lactobacillus and carnobacterium
Hammes, WALTER P and Hertel, CHRISTIAN
The Prokaryotes: Vol. 1: Symbiotic Associations, Biotechnology, Applied Microbiology, 4, 320-403 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service