Skip to Content
Merck
All Photos(2)

Key Documents

02382

Sigma-Aldrich

Methyltrioctylammonium hydrogen sulfate

greener alternative

≥95.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]3N(OSO3H)CH3
CAS Number:
Molecular Weight:
465.77
Beilstein:
7613669
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (T)

form

powder

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

61-65 °C

greener alternative category

SMILES string

OS([O-])(=O)=O.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

InChI

1S/C25H54N.H2O4S/c1-5-8-11-14-17-20-23-26(4,24-21-18-15-12-9-6-2)25-22-19-16-13-10-7-3;1-5(2,3)4/h5-25H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

MWSPFHZPVVWJCO-UHFFFAOYSA-M

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Phase transfer catalyst used with sodium tungstate dihydrate for greener oxidation under Noyori's conditions.

Noyori Oxidation (Sodium Tungstate Dihydrate, Hydrogen Peroxide, Methyltri-n-octylammonium Hydrogen Sulfate, Phosphonic Acid)

Other Notes

Catalyst for epoxidizing olefins with H2O2 in the absence of halogens

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marita Meurer et al.
Frontiers in microbiology, 10, 2911-2911 (2020-01-30)
Host defense peptides or antimicrobial peptides (AMPs), e.g., cathelicidins, have recently been discussed as a potential new treatment option against bacterial infections. To test the efficacy of AMPs, standardized methods that closely mimic the physiological conditions at the site of
Michelangelo Storari et al.
Food microbiology, 59, 213-223 (2016-07-05)
Clostridium tyrobutyricum is the main microorganism responsible for the late blowing defect in hard and semi-hard cheeses, causing considerable economic losses to the cheese industry. Deeper knowledge of the metabolic requirements of this microorganism can lead to the development of
S Van Vrekhem et al.
Scientific reports, 8(1), 4720-4720 (2018-03-20)
Insufficient glenoid fixation is one of the main reasons for failure in total shoulder arthroplasty. This is predominantly caused by the inert nature of the ultra-high molecular weight polyethylene (UHMWPE) used in the glenoid component of the implant, which makes
M J Hoes et al.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie, 19(5), 259-264 (1981-05-01)
The excretion of xanthurenic acid in urine after oral intake of 5 grams of L-tryptophan was studied and correlated with test and test-person parameters. The xanthurenic acid was determined by a colorimetric method, which gives reliable and reproducible results. Two
Bulletin of the Chemical Society of Japan, 70, 905-905 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service