Skip to Content
Merck
All Photos(1)

Key Documents

01302

Sigma-Aldrich

Fluosilicic acid

purum, 33.5-35%

Synonym(s):

Fluorosilicic acid, Hexafluorosilicic acid, Hydrogen hexafluorosilicate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SiF6
CAS Number:
Molecular Weight:
144.09
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38130308
PubChem Substance ID:
NACRES:
NA.21

grade

purum

Quality Level

concentration

33.5-35%

impurities

≤0.003% heavy metals (as Pb)

density

1.22 g/mL at 20 °C (lit.)
1.31 g/mL at 25 °C

anion traces

chloride (Cl-): ≤0.2%
phosphate (PO43-): ≤1000 mg/kg
sulfate (SO42-): ≤0.2%

cation traces

Fe: ≤100 mg/kg

SMILES string

F.F.F[Si](F)(F)F

InChI

1S/F4Si.2FH/c1-5(2,3)4;;/h;2*1H

InChI key

ZEFWRWWINDLIIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

High-silic fluoro silicic acids have been prepared. They have desired amount of extra silica upto 18% more than the composition as represented by H2SiF6. Preparation and properties of fluosilicic acid have been reported. Fluosilicic acid can be obtained as a major byproduct of phosphate fertilizer industry.It can be used in the preparation of sodium fluorosilicate, via neutralization with sodium chloride. It can also undergo neutralization with ammonia to produce ammonium fluoride and silica.

Application

Fluosilicic acid (FSA, H2SiF6) has been used in the following studies:
  • Fluoridation of fluoridated municipal water supplies.
  • Surface modification of calcium carbonate.
  • Liquid phase deposition (LPD) of silica.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Recovery of high surface area mesoporous silica from waste hexafluorosilicic acid (H 2 SiF 6) of fertilizer industry"
Sarawade.BP, et al.
Journal of Hazardous Materials, 173(01), 576-580 (2010)
Fluosilicic acid.
Journal of Chemical Education, 35(11), 562-562 (1958)
High-silica fluosilicic acids: Specific reactions and the equilibrium with silica.
Thomsen SM.
Journal of the American Chemical Society, 74(7), 1690-1693 (1952)
Surface modification of precipitated calcium carbonate using aqueous fluosilicic acid.
Lee CK.
Applied Surface Science, 202(1), 15-23 (2002)
Akash K Singh et al.
Aging cell, 21(9), e13675-e13675 (2022-08-14)
The master epigenetic regulator lysine acetyltransferase (KAT) p300/CBP plays a pivotal role in neuroplasticity and cognitive functions. Recent evidence has shown that in several neurodegenerative diseases, including Alzheimer's disease (AD), the expression level and function of p300/CBP are severely compromised

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service