Skip to Content
Merck
All Photos(1)

Documents

00071

Sigma-Aldrich

Acetaldehyde

ReagentPlus®, ≥99.0% (GC)

Synonym(s):

Ethanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHO
CAS Number:
Molecular Weight:
44.05
Beilstein:
505984
EC Number:
MDL number:
UNSPSC Code:
12352114
eCl@ss:
39021102
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

vapor density

1.52 (vs air)

vapor pressure

14.63 psi ( 20 °C)

product line

ReagentPlus®

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

365 °F

expl. lim.

60 %

refractive index

n20/D 1.332 (lit.)

bp

21 °C (lit.)

mp

−125 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC=O

InChI

1S/C2H4O/c1-2-3/h2H,1H3

InChI key

IKHGUXGNUITLKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetaldehyde is an organic colorless flammable liquid. It can be produced by hydration of acetylene and by catalytic oxidation of ethyl alcohol. It is mainly used in manufacturing acetic acid.
Mechanism of the thermal decomposition of acetaldehyde has been reported. It is a product of ethanol metabolism in the liver, binds covalently to various proteins, thereby alters the liver function and structure. The aldol condensation of acetaldehyde and heptanal in the presence of hydrotalcite-type catalysts has been described.

Application

Acetaldehyde may be used for the enzymatic synthesis of fatty acids.
It was used in the plasma polymerization for deposition of reactive aldehyde groups onto substrate, in a study to produce fibrous biomaterials with cell adhesive and also repulsive capability in biomedical applications. It was also used as calibration standard during determination of acetone, acetaldehyde, ethanol, and methanol in different human matrices, such as whole blood, vitreous humour, and urine using gas chromatography with flame ionization detection method (GC–FID).

Caution

forms easily paraldehyde

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-38.0 °F - closed cup

Flash Point(C)

-38.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

GC determination of acetone, acetaldehyde, ethanol, and methanol in biological matrices and cell culture.
Pontes, Helena, et al.
Journal of Chromatographic Science, 47.4, 272-278 (2009)
C S Lieber
Biochemical Society transactions, 16(3), 241-247 (1988-06-01)
Acetaldehyde, the toxic product of ethanol metabolism in the liver, covalently binds to a variety of proteins, thereby altering liver function and structure. Through its binding to tubulin, acetaldehyde decreases the polymerization of microtubules thereby impairing protein secretion and favouring
Bruce D. Craig, David S. Anderson
Handbook of Corrosion Data, 87-87 (1994)
Acetaldehyde plasma polymer-coated PET fibers for endothelial cell patterning: Chemical, topographical, and biological analysis.
Hadjizadeh, Afra.
Journal of Biomedical Materials Research. Part B, Applied Biomaterials, 94.1, 11-21 (2010)
The aldol condensation of acetaldehyde and heptanal on hydrotalcite-type catalysts.
Tichit D, et al.
J. Catal., 219(1), 167-175 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service