Skip to Content
Merck
All Photos(1)

Documents

00036

Supelco

ESI Tuning Mix, for Ion Trap

suitable for mass spectrometry (MS)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116004
NACRES:
NA.21

form

liquid

Quality Level

technique(s)

mass spectrometry (MS): suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Components

Analytes

  • Hexamethoxyphosphazene
  • Hexakis(2,2-difluoroethoxy)phosphazene
  • Hexakis(1H,1H,3H-tetrafluoropropoxy)phosphazene
  • Hexakis(1H,1H,5H-octafluoropentoxy)phosphazene
  • Hexakis(1H,1H,7H-dodecafluoroheptoxy)phosphazene
  • Hexakis(1H,1H,9H-perfluorononyloxy)phosphazene
  • 2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine
  • Betaine
  • Ammonium trifluoroacetate

Solvent composition (kg/kg):
  • Acetonitrile: 93.7%
  • Deionized water: 6.3%

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service