Skip to Content
Merck

Skip To

CHAPS-RO

Roche

CHAPS

98% (from N), suitable for dialysis, solubility: >50% (Aqueous solutions)

Synonym(s):

CHAPS, 3-[(3-cholamidopropyl)dimethyl-ammonio]-1-propane sulfonate, 3-[(3-cholamidopropyl)dimethyl-ammonio]-1-propane sulfonate, zwitterionic, Cholamidopropyl-dimethyl-ammonio]-1-propane sulfonate, 3-3-, zwitterionic, cholamidopropyl-dimethyl-ammonio]-1-propane sulfonate, 3-3-, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S
CAS Number:
Molecular Weight:
614.88
UNSPSC Code:
12161900
MDL number:
NACRES:
NA.54
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

zwitterionic

assay

98% (from N)

form

crystals

mol wt

micellar avg mol wt 6150, 614.88 g/mol (anhydrous basis)

packaging

pkg of 10 g (10810118001), pkg of 50 g (10810126001)

manufacturer/tradename

Roche

aggregation number

10

technique(s)

dialysis: suitable

CMC

4 mM, 6 mM (20-25°C)

transition temp

cloud point >100 °C

solubility

>50% (Aqueous solutions)

absorption

<0.06% at 278 nm in water at 1 % (w/v) (Ease of removal*: ++
Note:
* Arbitrary scale ranges from ++ to −)

storage temp.

2-8°C

SMILES string

C[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C32H58N2O7S/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41)/t21-,22?,23-,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

UMCMPZBLKLEWAF-RFCNGIAKSA-N

General description

3-[(3-Cholamidopropyl)dimethyl-ammonio]-1-propane sulfonate

Application

CHAPS is a nondenaturing zwitterionic detergent for membrane biochemistry.
Useful for solubilizing membrane proteins and breaking protein-protein interactions. CHAPS′ small micellar molecular weight (6,150) and high critical micelle concentration (6-10 mM) allow it to be removed from samples by dialysis. It is also suitable for protein solubilization for isoelectric focusing and two-dimensional electrophoresis. CHAPS is commonly used for non-denaturing (without urea) IEF and has been shown to give excellent resolution of some subcellular preparations and plant proteins. Concentrations between 2-4% (w/v) are typically used in an IEF gel.
Solubilization of membrane proteins. Combines the properties of both the sulfobetaine-type and bile-acid detergents.

Physical form

Crystals

Preparation Note

Working concentration: PAGE: 6.5 to 13 mM
Storage conditions (working solution): Do not store in solution, because precipitation may occur.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
3050-OP850500P19899
form

crystals

form

crystalline powder

form

powder

form

liquid

technique(s)

dialysis: suitable

technique(s)

-

technique(s)

protein quantification: suitable

technique(s)

protein quantification: suitable

assay

98% (from N)

assay

-

assay

>99% (TLC)

assay

-

aggregation number

10

aggregation number

10

aggregation number

-

aggregation number

-

storage temp.

2-8°C

storage temp.

10-30°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

>50% (Aqueous solutions)

solubility

-

solubility

-

solubility

-


Still not finding the right product?

Explore all of our products under CHAPS


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library