Skip to Content
Merck

Skip To

BSAV-RO

Roche

Bovine Serum Albumin

≥98.5%, New Zealand origin

Synonym(s):

Bovine Serum Albumin Fraction V

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.54
Biological source:
bovine
Assay:
≥98.5%
Form:
lyophilized
Technique(s):
ELISA: suitable, Southern blotting: suitable, western blot: suitable
Impurities:
<0.0003 Lithium (Flame photometry), <0.001 Iron (bathophenanthraline), <0.002 P (inorganic), <0.003 Heavy metals (as Pb), <0.003 Magnesium (xylidyl blue), <0.005 Glycerol (enzyme), <0.006 potassium (Flame photometry), <0.02 Calcium ( o-cresolphthalein), <0.05 glucose (enzyme), <0.1 L-lactate, <0.15 chloride (mercurom), <0.5 Sodium (flame photometry), <100 microorganisms (organism/g), <5 water
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

assay

≥98.5%

form

lyophilized

mol wt

Mr =68 kDa

packaging

pkg of 1 kg (10735108001), pkg of 100 g (10735086001), pkg of 50 g (10735078001), pkg of 500 g (10735094001)

manufacturer/tradename

Roche

origin

New Zealand origin

technique(s)

ELISA: suitable, Southern blotting: suitable, western blot: suitable

impurities

<0.0003 Lithium (Flame photometry), <0.001 Iron (bathophenanthraline), <0.002 P (inorganic), <0.003 Heavy metals (as Pb), <0.003 Magnesium (xylidyl blue), <0.005 Glycerol (enzyme), <0.006 potassium (Flame photometry), <0.02 Calcium ( o-cresolphthalein), <0.05 glucose (enzyme), <0.1 L-lactate, <0.15 chloride (mercurom), <0.5 Sodium (flame photometry), <100 microorganisms (organism/g), <5 water

absorption

≤0.4 at 550 nm

UniProt accession no.

foreign activity

Microorganisms <100 (Organism/g)

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

General description

For standard applications.

Application

Bovine Serum Albumin Fraction V is suitable for standard applications such as:
  • Stabilizer to prevent proteolysis and denaturation of enzymes and other proteins, especially when the proteins are present in low concentrations.[1]
  • Buffer component in immunochemistry, biochemistry, cell biology, or molecular biology
  • Reducing agent
  • Protein blocker for saturation of protein binding sites in ELISAs, Southern blots, and western blots[2]
  • Media supplement for media that require addition of protein
  • Carrier protein
  • Standard protein in gel electrophoresis and protein determination[3]

Preparation Note

Storage conditions (working solution): In solution, albumin is stable for about three months at -15 to -25 °C.

Analysis Note

Purity: ≤95% protein, ≤5% H2O; electrophoretically homogeneous
Contaminants: <0.5% Na (flame photometric), <0.006% K (flame photometric), <0.0003% Li (flame photometric), <0.02% Ca (o-cresolphthalein), <0.003% Mg (xylidyl blue), <0.003% heavy metals (as Pb), <0.001% Fe (bathophenanthroline), <0.002% Pi, <0.05% glucose (enzymatic), <0.005% glycerol (enzymatic), <0.025% L-lactate (enzymatic)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
81053381053N810535
assay

≥98.5%

assay

≥98% (purity)

assay

≥98% (purity)

assay

≥98% (purity)

technique(s)

ELISA: suitable, western blot: suitable, Southern blotting: suitable

technique(s)

blood typing: suitable ( )

technique(s)

blood typing: suitable ( )

technique(s)

blood typing: suitable ( )

biological source

bovine

biological source

bovine serum

biological source

bovine serum

biological source

bovine serum

origin

New Zealand origin

origin

USA origin

origin

USA origin

origin

USA origin

form

lyophilized

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

Gene Information

bovine ... ALB(280717)

Gene Information

-

Gene Information

-

Gene Information

-


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Laurent Berchebru et al.
Journal of microbiological methods, 105, 141-145 (2014-07-20)
Test and evaluation of engineered biothreat agent detection systems ("biodetectors") are a challenging task for government agencies and industries involved in biosecurity and biodefense programs. In addition to user friendly features, biodetectors need to perform both highly sensitive and specific
Leonie Kollenstart et al.
Scientific reports, 11(1), 12795-12795 (2021-06-19)
The collection of known posttranslational modifications (PTMs) has expanded rapidly with the identification of various non-acetyl histone lysine acylations, such as crotonylation, succinylation and butyrylation, yet their regulation is still not fully understood. Through an unbiased chromatin immunoprecipitation (ChIP)-based approach
Paulina Krzysica et al.
Animals : an open access journal from MDPI, 12(21) (2022-11-12)
Cytokines like interferon (IFN)-γ, interleukin (IL)-2, IL-6, IL-10, and IL-12p40 are important biomarkers for characterizing the nature and strength of immune responses. It is important to be able to quantify the cytokines at the protein level in biological samples. Quantification



Global Trade Item Number

SKUGTIN
1073509400104061838258885
1073508600104061838258878
1073507800104061838258861
1073510800104061838674852