Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bovine Serum Albumin
Synonym(s):
Bovine Serum Albumin
Products
Product Category
albumins (3)
Brand
Sigma-Aldrich (1)
Biological Source
Manufacturer
Physical Form
Quality Segment
Geographic Origin
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
BSAV-RO
Bovine Serum Albumin,
≥98.5%, New Zealand origin
Expand
View Pricing
10711454001
Bovine Serum Albumin,
blood typing: suitable, Endonuclease, Exonuclease, Rnases, Proteinases free
Expand
View Pricing
10775835001
Bovine Serum Albumin,
98.5% (electrophoresis), Fatty acids (total) ≤0.2 mg/g, Triglycerides (enzym.) free, Immunoglobulins not detectable, microbiological culture: suitable, USA origin
Expand
View Pricing
20-191
Bovine Serum Albumin,
10% in TBS
Expand
View Pricing