Skip to Content
Merck
All Photos(1)

Key Documents

11140965001

Roche

ATP

Lithium salt

Synonym(s):

lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

Quality Level

Assay

97% (HPLC)

form

solution

packaging

pkg of 400 μL (4,000 cm2 membrane)

manufacturer/tradename

Roche

concentration

100 mM

storage temp.

−20°C

General description

ATP is a 100mM solution of the lithium salt (pH 7).

Application

ATP, lithium salt has been used as a component in:
  • loading buffer
  • in vitro transcription reaction mixture
  • ligation mix

Biochem/physiol Actions

Adenosine triphosphate (ATP) is a central metabolite, which acts as an energy transfer molecule, a phosphate donor and a signaling molecule inside the cells. This extracellular signaling molecule also serves as a neurotransmitter in peripheral and central nervous systems. In the peripheral nervous system, ATP participates in chemical transmission.

Components

Contents
ATP, lithium salt, 100mM

Quality

Typical analysis:
97% ATP (HPLC), ≤1.5% ADP (HPLC), ≤0.5% AMP (HPLC)
Tested for the absence of RNases according to current Quality Control procedures
Updated contents under Features & Benefits on CorpBase

Unit Definition

1mM ATP = 15.3A260 units

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imaging changes in the cytosolic ATP-to-ADP ratio
Methods in Enzymology, 547, 355-371 (2014)
Simultaneous generation of many RNA-seq libraries in a single reaction
Shishkin AA, et al.
Nature Methods, 12(4), 323-323 (2015)
Anna Wypijewska Del Nogal et al.
International journal of molecular sciences, 22(16) (2021-08-28)
Pre-miRNA-377 is a hairpin-shaped regulatory RNA associated with heart failure. Here, we use single-molecule optical tweezers to unzip pre-miRNA-377 and study its stability and dynamics. We show that magnesium ions have a strong stabilizing effect, and that sodium ions stabilize
Imaging changes in the cytosolic ATP-to-ADP ratio
Test, 547, 355-371 (2014)
Oxidized low-density lipoprotein is a novel predictor of interferon responsiveness in chronic hepatitis C infection
Solbach P, et al.
Cellular and molecular gastroenterology and hepatology, 1(3), 285-294 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service