Skip to Content
Merck
All Photos(1)

Key Documents

11093266910

Roche

Streptavidin-AP-conjugate for nucleic acid detection

Synonym(s):

streptavidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106

form

solution

Quality Level

packaging

pkg of 200 μL (150U)

manufacturer/tradename

Roche

storage temp.

2-8°C

Application

For the detection of biotin-labeled nucleic acids.

Physical form

Solution in 50 mM triethanolamine buffer, 3M NaCl, 1 mM MgCl2, 0.1 mM ZnCl2, 10 mg/ml bovine serum albumin, pH 7.6.

Preparation Note

Working Solution:
Prepare by diluting the conjugate solution 1:5,000 with 100 mM Tris-HCl, 150mM NaCl, pH 7.5

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Rodemer et al.
Frontiers in cellular neuroscience, 14, 61-61 (2020-04-09)
Traumatic spinal cord injury (SCI) results in persistent functional deficits due to the lack of axon regeneration within the mammalian CNS. After SCI, chondroitin sulfate proteoglycans (CSPGs) inhibit axon regrowth via putative interactions with the LAR-family protein tyrosine phosphatases, PTPσ
Sumonto Mitra et al.
Frontiers in aging neuroscience, 13, 714186-714186 (2021-09-04)
Gradual decline in cholinergic transmission and cognitive function occurs during normal aging, whereas pathological loss of cholinergic function is a hallmark of different types of dementia, including Alzheimer's disease (AD), Lewy body dementia (LBD), and Parkinson's disease dementia (PDD). Glial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service