RABHRP3
HRP-Streptavidin
Synonym(s):
HRP streptavidin, HRP-conjugated streptavidin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
shipped in
wet ice
storage temp.
−20°C
General description
Streptavidin is a biotin-binding protein. Horseradish peroxidase (HRP) is a heme-containing enzyme. HRP-Streptavidin consists of the protein streptavidin, as a secondary detection reagent, conjugated to the enzyme HRP. The conjugate is used in ELISA to increase the detection of various kinds of targets.
Application
HRP-Streptavidin has been conjugated to secondary antibodies for western blotting.
Other Notes
This item is for RAB series EIA kits.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Archives of virology, 165(3), 627-641 (2020-01-23)
HCV is a potential cause of viral hepatitis, which leads to blood-borne chronic hepatitis, liver cirrhosis and hepatocellular carcinoma. Anti-HCV antibody detection assays detect HCV infection after approximately 70 days. HCV core antigen can be detected much earlier than anti-HCV
Functional Protein Microarrays in Drug Discovery (2007)
Electrophoresis, 42(6), 793-799 (2020-12-24)
Western blot (protein immunoblot) is a widely used analytical technique in molecular biology. Utilizing the specific recognizing primary antibody, proteins immobilized on various matrix are investigated by subsequent visualization steps, for example, by the horse radish peroxidase conjugated secondary antibody
Biotin-Streptavidin Competition Mediates Sensitive Detection of Biomolecules in Enzyme Linked Immunosorbent Assay
PLoS ONE, 11(3), e0151153-e0151153 (2016)
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 240, 110591-110591 (2019-11-02)
The purpose of this study was to investigate changes in expression of known cellular regulators of metabolism during hyperphagia (Sept) and hibernation (Jan) in skeletal muscle and adipose tissue of brown bears and determine whether signaling molecules and transcription factors
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service