Skip to Content
Merck
All Photos(1)

Key Documents

10102547001

Roche

AMP-PNP

Adenylyl-imidodiphosphate

Synonym(s):

Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate, β,γ-Imidoadenosine 5′-triphosphate lithium salt hydrate, AMP-PNP, ATP[β,γ-NH], Adenylyl imidodiphosphate lithium salt hydrate, App(NH)p

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N6O12P3 · xLi+ · yH2O
CAS Number:
Molecular Weight:
506.20 (free acid basis)
Beilstein:
6047109
UNSPSC Code:
12352204

form

powder

mol wt

Mr 506.2 (ANP-PNP)
Mr 529.9 (AMP-PNP-Li4)

packaging

pkg of 25 mg

manufacturer/tradename

Roche

SMILES string

[Li+].[Li+].[Li+].[Li+].[H]O[H].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)NP([O-])([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C10H17N6O12P3/c11-8-5-9(13-2-12-8)16(3-14-5)10-7(18)6(17)4(27-10)1-26-31(24,25)28-30(22,23)15-29(19,20)21/h2-4,6-7,10,17-18H,1H2,(H,24,25)(H2,11,12,13)(H4,15,19,20,21,22,23)/t4-,6-,7-,10-/m1/s1

InChI key

PVKSNHVPLWYQGJ-KQYNXXCUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

AMP-PNP (adenylyl-imidodiphosphate) is a non-hydrolysable analogue of ATP. It inhibits fast axonal transport and facilitates the interaction between membranous organelles and microtubules.
AMP-PNP(adenylyl-imidodiphosphate) is an artificial substrate of enzyme, adenylate cyclase. Adenylate cyclase helps to hydrolyze adenylyl-imidodiphosphate slowly than ATP.

Application

AMP-PNP has been used in the preparation of nucleotide solutions, two-motor fluorescence assay, preparation of flow-cells with kinesin and cascade binding assays.
AMP-PNP is a competitive inhibitor of most ATP-dependent systems. A good substrate for enzymes hydrolyzing between the α- and β-phosphorus atom, yet is resistant to enzymes cleaving between the β- and γ-phosphorus atom. Substrate of snake venom phosphodiesterase and adenylate cyclase.
AMP-PNP has been used for two-motor fluorescence assay.

Features and Benefits

Contents
88% AMP-PNP (from N), 4.5% lithium, 6% water

Quality

Contaminants: ≤1.5% Pi , <0.1% ATP (enzymatically)

Storage and Stability

Store at -15–-25 °C. (Store dry!)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High accuracy measurements of nanometer-scale distances between fluorophores at the single-molecule level
Niekamp S, et al.
bioRxiv, 234740-234740 (2017)
Nucleotide dependence of the dimerization of ATP binding cassette nucleotide binding domains
Fendley GA, et al.
Biochemical and Biophysical Research Communications, 480(2), 268-272 (2016)
Arthur Chow et al.
Methods in enzymology, 667, 365-402 (2022-05-08)
Pseudokinases often operate through functionally related enzymes and receptors. A prime example is the pseudokinase KSR (Kinase Suppressor of RAS), which can act as both an amplifier and inhibitor of members in the RAS-MAPK (Mitogen Activated Protein Kinase) signaling pathway.
Cristina Olivieri et al.
eLife, 9 (2020-04-28)
In the nucleus, the spatiotemporal regulation of the catalytic subunit of cAMP-dependent protein kinase A (PKA-C) is orchestrated by an intrinsically disordered protein kinase inhibitor, PKI, which recruits the CRM1/RanGTP nuclear exporting complex. How the PKA-C/PKI complex assembles and recognizes
Maximilian M Biebl et al.
Nature communications, 12(1), 828-828 (2021-02-07)
The co-chaperone p23 is a central part of the Hsp90 machinery. It stabilizes the closed conformation of Hsp90, inhibits its ATPase and is important for client maturation. Yet, how this is achieved has remained enigmatic. Here, we show that a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service