ZAFS10EWW
AFS® Water Purification System
Water purification system with Elix® technology, producing CLRW quality water up to 200 L/day
Synonym(s):
AFS® 10E Water Purification System
About This Item
Recommended Products
AC/DC input
100 - 230 V, 50 - 60 Hz
Quality Level
product line
AFS®
packaging
pkg of 1 unit
parameter
10 L/hr make-up flow rate
2 L/min distribution flow rate
2 L/min product water instant delivery rate
200 L/day max. usage
28 kg operating weight (61.7 lb)
resistivity
>15 MΩ-cm, at 25°C
system H × W × depth
58.5 cm (23 in.) × 26.8 cm (10.6 in.) × 42.6 cm (16.8 in.)
total organic carbon (TOC) residue
<30 ppb
input
potable tap water
output
clinical laboratory reagent water (CLRW) (that complies with the CLSI guideline)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- State-of-the-art Elix® electrodeionization (EDI) technology is combined with other complementary water purification techniques to provide consistent water quality meeting CLRW standards
- Elix® EDI technology uses permanently self-regenerating ion exchange resins to ensure predictable running costs
- Fulfills accreditation needs (e.g. CAP 15189SM accreditation to the ISO 15189:2022 standard) with full monitoring and automatic water quality archiving capabilities. Up to 6 months of information can be stored for reliable traceability
- Patented Elix® technology provides constant quality water without the need for softeners or conditioning cartridges, which means reduced maintenance ― and less analyzer downtime
- Milli-Q® Service Plans offer a range of support, including preventive maintenance visits and online contract management
- A small footprint allows installation wherever it’s convenient: on the wall, on or under the bench, or on a cart
Components
- The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
- This catalog number does not include consumables, accessories and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
- Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!
Other Notes
- Organism Retention: Microorganisms
- Mode of Action: Filtering and UV
- Application: Clinical laboratories
- Intended Use: Water purification
- Instructions for Use: This item provides water filtered through 0.22 ?m sterilizing-grade membranes and exposed to bactericidal UV lamp. Please refer to system equipment user guide section "System" (provided on USB key in the North American configuration version).
- Storage Statement: Store in dry location
- Disposal Statement: Dispose of in accordance with applicable federal, state, and local regulations.
Legal Information
Disclaimer

Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service