Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
AFS
®
Water Purification System
Products
Product Category
water purification systems (7)
Brand
Milli-Q (7)
Product Line
Quality Segment
Daily Water Usage (L/day)
Package Size
Output Water Quality
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
ZAFS15EWW
AFS
®
Water Purification System,
Water purification system with Elix
®
technology, producing CLRW quality water up to 300 L/day
Expand
View Pricing
ZAFS08D0WW
AFS
®
Water Purification System,
For analyzers with degassed pure water needs up to 160 L/day
Expand
View Pricing
ZAFS10EWW
AFS
®
Water Purification System,
Water purification system with Elix
®
technology, producing CLRW quality water up to 200 L/day
Expand
View Pricing
ZAFS0080WW
AFS
®
Water Purification System,
For analyzers with pure water needs up to 160 L/day
Expand
View Pricing
ZAFS0160WW
AFS
®
Water Purification System,
For analyzers with pure water needs up to 320 L/day
Expand
View Pricing
ZAFS0240WW
AFS
®
Water Purification System,
For analyzers with pure water needs up to 480 L/day
Expand
View Pricing
ZAFS16D0WW
AFS
®
Water Purification System,
For analyzers with degassed pure water needs up to 320 L/day
Expand
View Pricing