Skip to Content
Merck
All Photos(2)

Key Documents

SCC065

Sigma-Aldrich

HL-1 Cardiac Muscle Cell Line

Mouse

Synonym(s):

HL-1 Cardiomyocyte Cell Line, HL-1 Atrial Muscle Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

Product Name

HL-1 Cardiac Muscle Cell Line, HL-1 Cardiac Muscle Cell Line has been extensively characterized and is a valuable model system to address questions of cardiac biology at the cellular & molecular levels.

biological source

mouse

Quality Level

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable

General description

HL-1 Cardiac Muscle Cell Line is an immortalized mouse cardiomyocyte cell line able to continuously divide and spontaneously contract while maintaining a differentiated cardiac phenotype. HL-1 can be serially passaged without losing its differentiated cardiac myocyte phenotype, including morphological, biochemical, and electrophysiological properties. HL-1 cells have been used to study normal cardiomyocyte function with regard to signaling, electrical, metabolic, and transcriptional regulation as well as to address pathological conditions such as hypoxia, hyperglycemia-hyperinsulinemia, apoptosis, and ischemia-reperfusion.

The HL-1 line was derived from AT-1 subcutaneous tumor excised from an adult female C57BL/6J mouse. The parental AT-1 line was originally derived from an atrial tumor growing in a transgenic mouse in which expression of the SV40 large T-antigen was targeted to atrial cardiomyocytes via the atrial natriuretic factor (ANF) promoter.

Cell Line Description

Cardiomyocyte Cells

Application

Research Category
Cardiac

Cardiovascular Disease

Toxicity
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells tested negative for infectious diseases by the Mouse Essential CLEAR panel PCR panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination.
• Functional Assay: Fluo-8 calcium dye fluorescent based assay for detecting changes in intracellular calcium

Storage and Stability

HL-1 Cardiac Muscle Cell Line should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hailong Cao et al.
American journal of translational research, 12(9), 5032-5047 (2020-10-13)
Persistent atrial fibrillation (PeAF) is a progressive cardiovascular disease with a high risk for most patients after diagnosis. Poor molecular description of PeAF has led to unsatisfactory interpretation of the pathogenesis of it, resulting in the lack of effective treatments.
W C Claycomb et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(6), 2979-2984 (1998-04-18)
We have derived a cardiac muscle cell line, designated HL-1, from the AT-1 mouse atrial cardiomyocyte tumor lineage. HL-1 cells can be serially passaged, yet they maintain the ability to contract and retain differentiated cardiac morphological, biochemical, and electrophysiological properties.
Jian Hou et al.
Journal of thoracic disease, 12(5), 2039-2048 (2020-07-10)
Atrial fibrillation (AF) is the most common sustained arrhythmia. DACT2 is a novel and important mediator of signaling pathways. The aim of this study was to investigate the clinical significance and functions of DACT2 expression in AF. Immunohistochemistry was used
Paola Lagonegro et al.
Nature communications, 13(1), 6-6 (2022-01-12)
Myocardial infarction causes 7.3 million deaths worldwide, mostly for fibrillation that electrically originates from the damaged areas of the left ventricle. Conventional cardiac bypass graft and percutaneous coronary interventions allow reperfusion of the downstream tissue but do not counteract the
Steven M White et al.
American journal of physiology. Heart and circulatory physiology, 286(3), H823-H829 (2004-02-10)
HL-1 cells are currently the only cardiomyocyte cell line available that continuously divides and spontaneously contracts while maintaining a differentiated cardiac phenotype. Extensive characterization using microscopic, genetic, immunohistochemical, electrophysiological, and pharmacological techniques has demonstrated how similar HL-1 cells are to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service