Skip to Content
Merck
All Photos(1)

Key Documents

QIA58

Sigma-Aldrich

BrdU Cell Proliferation Assay

This proliferation assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells.

Synonym(s):

Bromodeoxyuridine Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.84

Quality Level

usage

sufficient for 1000 tests
sufficient for 200 tests

packaging

pkg of 1 96-well plate(s)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

input

sample type intact cells
sample type intact cells

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

General description

A non-isotopic enzyme immunoassay for the quantification of cell proliferation.Evaluation of cell cycle progression is essential for investigations in many scientific fields. Measurement of [3H] thymidine incorporation as cells enter S phase has long been the traditional method for the detection of cell proliferation. Subsequent quantification of [3H] thymidine is performed by scintillation counting or autoradiography. This technology is slow, labor intensive and has several limitations including the handling and disposal of radioisotopes and the necessity of expensive equipment. A well-established alternative to [3H] thymidine uptake has been demonstrated by numerous investigators. In these methods bromodeoxyuridine (BrdU), a thymidine analog replaces [3H] thymidine. BrdU is incorporated, into newly synthesized DNA strands of actively proliferating cells. Following partial denaturation of double stranded DNA, BrdU is detected immunochemically allowing the assessment of the population of cells, which are actively synthesizing DNA. The Calbiochem® BrdU Cell Proliferation Assay involves incorporation of BrdU into cells cultured in plates and BrdU immunolabeling using the cell layer as the solid phase. The resultant assay is sensitive, rapid, easy to perform and applicable to high sample throughput. In addition to evaluation of cell proliferation, information such as cell number, morphology and analysis of cellular antigens can be obtained from a single culture.
BrdU Cell Proliferation Assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells. It is sensitive, rapid, and easy to perform.
Note: 1 T = 1 test.
This proliferation assay is a non-isotopic immunoassay for quantification of BrdU incorporation into newly synthesized DNA of actively proliferating cells.

Components

BrdU Label, Fixative/Denaturing Solution, BrdU Antibody, Antibody Diluent, Peroxidase Goat Anti-Mouse IgG, Conjugate Diluent, Substrate, Plate Wash Concentrate, Stop Solution, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Specifications

Assay Time: 3 h

Principle

The Calbiochem® BrdU Cell Proliferation Assay is a non-isotopic immunoassay for the quantitation of bromodeoxyuridine incorporation into newly synthesized DNA of actively proliferating cells.

Storage and Stability

Upon arrival store the entire contents of the kit at -20°C, in a non-frostfree freezer.



Prior to use:

1. Remove the Fixative/Denaturing Solution and place at room temperature for at least 4 h prior to use. Precipitates that may occur while cold should go back into solution.

2. Reconstitute the Peroxidase Goat Anti-Mouse IgG with the appropriate volume of 1X PBS.

• For the 200 test kit, reconstitute Peroxidase Goat Anti-Mouse IgG in 125 µl of 1X PBS. Use Conjugate Diluent for any further lot-specific dilution.

• For the 1000 test kit, reconstitute Peroxidase Goat Anti-Mouse IgG in 250 µl of 1X PBS. Use Conjugate Diluent for any further lot-specific dilution.

Allow solution to stand at room temperature for 10 min. (or until solution is clear). Divide into small aliquots; aliquots not to be used that same day should be stored at -20°C in a non-frostfree freezer.



Once the kit components have been thawed for use:

• The BrdU label and 100X Anti-BrdU Antibody should be divided into small aliquots and stored at -20°C in a non-frostfree freezer with the Peroxidase Goat Anti-Mouse IgG; avoid multiple freeze/thaw cycles.

• All the other kit components (Substrate, Antibody Diluent, Conjugate Diluent, 20X Plate Wash Concentrate, Fixative/Denaturing Solution, Stop Solution) should be stored at 4°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Hardonk, M.J. and Harms, G. 1990. Acta histochemica, Suppl.39, 99.
Muir, D., et al. 1990. Analytical Biochemistry185, 377.
Lanier, T. L., et al. 1989. Carcinogenesis10, 1341.
Magaud, J.-P., et al. 1988. J. Immunol. Methods106, 95.
Collins, S.J. 1987. Blood70, 1233.
Porstmann, T., et al. 1985. J. Immunol. Methods82, 169.
Raza, A., et al. 1985. Cytometry6, 633.
Morstyn, G., et al. 1983. J. Clin. Invest.72, 1844.
Gratzner, H.G. 1982. Science218, 474.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anika Langenfurth et al.
Scientific reports, 6, 32203-32203 (2016-08-31)
The subventricular zone (SVZ) provides a constant supply of new neurons to the olfactory bulb (OB). Different studies have investigated the role of olfactory sensory input to neural precursor cell (NPC) turnover in the SVZ but it was not addressed
Jin Dai et al.
International journal of biological sciences, 9(10), 1089-1098 (2013-12-18)
To investigate the effects of Genistein on the osteogenic related gene expression profiles during osteoblastic differentiation of human bone marrow mesenchymal stem cell (hBMSC) cultures, the hBMSCs were cultured under osteogenic differentiation medium with the addition of Genistein (10(-8)∼10(-5) M)
Haiyan Ge et al.
PloS one, 7(12), e51821-e51821 (2012-12-29)
Dexamethasone (DEX) co-treatment has proved beneficial in NSCLC patients, improving clinical symptoms by the reduction of side effects after chemotherapy. However, recent studies have shown that DEX could render cancer cells more insensitive to cytotoxic drug therapy, but it is
Hyun Jun Jung et al.
PloS one, 6(12), e28492-e28492 (2011-12-07)
Aquaporin (AQP) is a family of transmembrane proteins for water transport. Recent studies revealed that AQPs are likely to play a role in tumor progression and invasion. We aimed to examine the potential role of AQP5 in the progression of
Zhousheng Xiao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(7), 2418-2432 (2011-04-02)
We investigated whether polycystin-1 is a bone mechanosensor. We conditionally deleted Pkd1 in mature osteoblasts/osteocytes by crossing Dmp1-Cre with Pkd1(flox/m1Bei) mice, in which the m1Bei allele is nonfunctional. We assessed in wild-type and Pkd1-deficient mice the response to mechanical loading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service