Skip to Content
Merck
All Photos(1)

Key Documents

PC548

Sigma-Aldrich

Anti-Porin (Ab-5) (185-197) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-VDAC (Voltage-Dependent Anion Channel)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VDAC1(7416)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~31 kDa (VDAC) porin protein.
Recognizes the ~31 kDa porin protein.
This Anti-Porin (Ab-5) (185-197) Rabbit pAb is validated for use in Immunoblotting for the detection of Porin (Ab-5) (185-197).

Immunogen

Human
a synthetic peptide corresponding to amino acids 185-197 of human porin

Application


Immunoblotting (2 g/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 1 mg/ml BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Rat heart tissue

Other Notes

Crompton, M., et al. 1999. Biochem. J.341, 233.
Yu, W.H., et al. 1995. J. Biol. Chem.270, 13998.
McEnry, M.W., et al. 1993. J. Biol. Chem.268, 23289.
The immunogen peptide is 100% conserved in all three isoforms of VDAC (VDAC1, VDAC2, and VDAC3). Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Ishiwata et al.
PloS one, 7(8), e42118-e42118 (2012-08-08)
CryAB (HspB5) and HspB2, two small heat shock genes located adjacently in the vertebrate genome, are hypothesized to play distinct roles. Mice lacking both cryab and hspb2 (DKO) are viable and exhibit adult-onset degeneration of skeletal muscle but confounding results
Theresa Schöttl et al.
Molecular metabolism, 4(9), 631-642 (2015-09-29)
Several human and rodent obesity studies speculate on a causal link between altered white adipocyte mitochondria in the obese state and changes in glucose homeostasis. We here aimed to dissect whether alterations in white adipocyte mitochondrial respiratory function are a
Xiaoxiao Peng et al.
Scientific reports, 6, 28080-28080 (2016-06-28)
Recently thioredoxin reductase 1 (TrxR1), encoded by Txnrd1, was suggested to modulate glucose and lipid metabolism in mice. Here we discovered that TrxR1 suppresses insulin responsiveness, anabolic metabolism and adipocyte differentiation. Immortalized mouse embryonic fibroblasts (MEFs) lacking Txnrd1 (Txnrd1(-/-)) displayed
Rianne Nederlof et al.
BMC anesthesiology, 17(1), 51-51 (2017-03-31)
Remote ischemic preconditioning (RIPC) efficacy is debated. Possibly, because propofol, which has a RIPC-inhibiting action, is used in most RIPC trials. It has been suggested that clinical efficacy is, however, present with volatile anesthesia in the absence of propofol, although
Rianne Nederlof et al.
Scientific reports, 7(1), 12749-12749 (2017-10-08)
Both the absence of cyclophilin D (CypD) and the presence of mitochondrial bound hexokinase II (mtHKII) protect the heart against ischemia/reperfusion (I/R) injury. It is unknown whether CypD determines the amount of mtHKII in the heart. We examined whether CypD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service