GF316
EGF Protein, Human Recombinant Animal Free
This Recombinant animal free human EGF is manufactured using all non-animal reagents.
Synonym(s):
Epidermal Growth Factor, Urogastrone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
recombinant
expressed in E. coli
Quality Level
Assay
>98% (SDS-PAGE and HPLC)
impurities
<0.1 μg/mg protein endotoxin (<1EU/μg)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
General description
Epidermal growth factor (EGF) is the founding member of the EGF family that also includes TGFα, amphiregulin (AR), betacellulin (BTC), epiregulin (EPR), heparinbinding EGF like growth factor (HB-EGF), epigen, and the neuregulins (NRG)1-6. EGF is a potent growth factor that stimulates the proliferation of various epidermal and epithelial cells. Additionally, EGF has been shown to inhibit gastric secretion, and to be involved in wound healing. EGF signals through a receptor known as c-erbB, which is a class I tyrosine kinase receptor. This receptor also binds with TGF-α and VGF (vaccinia virus growth factor). Recombinant human EGF is a 6.2 kDa globular protein containing 53 amino acid residues including 3 intramolecular disulfide-bonds.
Product Source: E. coli
Specificity
Cross Reactivty
Cow, Hamster, Monkey, Mouse, Pig, Rabbit, Rat
Cow, Hamster, Monkey, Mouse, Pig, Rabbit, Rat
Application
Research Category
Stem Cell Research
Stem Cell Research
Research Sub Category
Growth Factors & Receptors
Growth Factors & Receptors
Quality
Bioactivity assay: The ED50 was determined by a cell proliferation assay using balb/c 3T3 cells is ≤0.1 ng/mL, corresponding to a specific activity of ≥1 x 107 units/mg.
Physical form
Product is filtered through a 0.2 micron filter before lyophilization.
Recombinant animal free human EGF is manufactured using all non-animal reagents
Storage and Stability
Store at -20°C for up to 6 months from date of receipt.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service