MCFLX4702
MILLICUP-FLEX, 47mm Disposable Filtration Kit; Nonsterile, Solvent-Resistant; Semi-reusable support collars
Great for Mobile Phase - 250mL Disposable Vacuum Filtration Unit w/ GL45 Threaded Connection; Compatible with 47mm membranes; Nonsterile Semi-reusable: 10 uses/kit
Synonym(s):
47mm Filtration Kit, MILLICUP-FLEX Filtration Kit, Nonsterile Filtration Kit
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41101708
NACRES:
NB.24
Recommended Products
material
polypropylene device
sterility
non-sterile
technique(s)
vacuum filtration: suitable
filter capacity
250 mL
funnel capacity
250 mL
fitting
inlet funnel
outlet bottle top
application(s)
filtration
Related Categories
General description
The Millicup-FLEX nonsterile, vacuum-driven filtration unit combines the convenience of disposable filtration with the flexibility and compatibility of traditional vacuum filtration glassware. With solvent-resistant, modular components, the Millicup-FLEX filtration unit offers a safe and efficient means for filtering a variety of aqueous and organic solutions, allowing the user to filter directly into GL45 vacuum-rated filtration bottles with their choice of 47mm disc membranes (sold separately). The ready-to-assemble, clampless design of the Millicup-FLEX filtration system eliminates the need for cleaning prior to filtration – saving time and ultimately reducing the risk of sample contamination. Following filtration, the 250mL funnel and membrane support collar easily disconnect to enable direct access to the membrane.
Features & Benefits:
Available in 2 versions:
All three components of the Millicup-FLEX filtration unit are fully recyclable, minimizing your environmental impact.
Features & Benefits:
- Compatible with organic and aqueous solvents
- Ergonomic, clampless design
- Reduce contamination risk
- Filter directly into vacuum-rated storage bottles
- Easy access to membrane after filtration (membranes sold separately)
- Fully recyclable components
Available in 2 versions:
- Semi-reusable Kit: 10 funnels with 2 semi-reusable membrane support collars and vacuum tube adapters (MCFLX4702)
- Single-Use Kit: 10 funnels with 10 single-use membrane support collars and vacuum tube adapters (MCFLX4710)
All three components of the Millicup-FLEX filtration unit are fully recyclable, minimizing your environmental impact.
Application
- Laboratory Filtration
- Air/gas/organic solvent filtration and venting
- Clarification of water, buffers or aqueous solutions
- Environmental Analysis
- Food and Beverage
- Industrial QC
- Pharmaceutical Products Analysis
- Water Monitoring
Direct patient care: No
Components
Kit includes: 10 funnels, 2 support collars, and 2 vacuum tube adapters
Principle
Vacuum
Physical form
Color code: Clear with blue adapter
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service