58062-U
Supelco® Mobile Phase Filtration Apparatus 2
Synonym(s):
Supelco Apparatus for Mobile Phase Filtration
All Photos(1)
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
Recommended Products
description
connects to aspiration line yes
packaging
pkg of 1
General description
Connects to aspiration line
Includes:
Includes:
- 250 mL glass reservoir
- S/T 34/45 funnel base
- S/T 34/45 1000 mL flask and glass cap
- Clamp
- Stainless steel holder and screen
- 10 PTFE gaskets
- 50 Nylon 66 filters (47 mm, 0.45 μm pores).
Legal Information
Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)